Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00657677_mH
          See other HSD3B5 GE Assays ›
          Gene Symbol:
          Hsd3b5
          Gene Name:
          hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 98618636 - 98645534 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00657677_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hsd3b5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008295.2 NP_032321.2 2 - 3 259 87
          GenBank mRNA BC012715.1 - 2 - 3 263 4193717 87
          L41519.1 - 2 - 3 244 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002554.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Mouse Androgen Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hsd3b5

          Panther Classification:

          Molecular Function -

          dehydratase

          Gene Ontology Categories:

          Function(s) Process(es)

          steroid biosynthetic process
          oxidation-reduction process
          C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity
          3-beta-hydroxy-delta5-steroid dehydrogenase activity
          aldo-keto reductase (NADP) activity
          isocitrate dehydrogenase activity
          mevaldate reductase activity
          steroid binding
          gluconate dehydrogenase activity
          steroid dehydrogenase activity
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          epoxide dehydrogenase activity
          5-exo-hydroxycamphor dehydrogenase activity
          2-hydroxytetrahydrofuran dehydrogenase activity
          acetoin dehydrogenase activity
          phenylcoumaran benzylic ether reductase activity
          D-xylose:NADP reductase activity
          L-arabinose:NADP reductase activity
          D-arabinitol dehydrogenase, D-ribulose forming (NADP+) activity
          steroid dehydrogenase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
          steroid dehydrogenase activity, acting on the CH-CH group of donors
          (R)-(-)-1,2,3,4-tetrahydronaphthol dehydrogenase activity
          3-hydroxymenthone dehydrogenase activity
          very long-chain-3-hydroxyacyl-CoA dehydrogenase activity
          dihydrotestosterone 17-beta-dehydrogenase activity
          (R)-2-hydroxyisocaproate dehydrogenase activity
          L-arabinose 1-dehydrogenase (NADP+) activity
          L-xylulose reductase (NAD+) activity
          3-ketoglucose-reductase activity
          NAD binding
          (R)-2-hydroxyglutarate dehydrogenase activity
          D-arabinitol dehydrogenase, D-xylulose forming (NADP+) activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111980 BY460328.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111980 L41519.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111980 NM_008295.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111980 BC012715.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397045 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 437950 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 828418 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 642598 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 137306 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 115586 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 96403 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122447 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111980 BY460328.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111980 L41519.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111980 NM_008295.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111980 BC012715.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P496162 BY460328.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P496162 L41519.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P496162 NM_008295.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P496162 BC012715.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05002668 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05002678 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122447 - -
          Whole Mouse Genome G4122A A_51_P496162 BY460328.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P496162 L41519.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P496162 NM_008295.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P496162 BC012715.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131445 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05002678 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122447 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02592962 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02932066 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02932076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02880137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03064622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02437457 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02585211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22360415 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10500537 - -
          Affymetrix Mouse Gene 1.0 ST Array 10500540 - -
          Affymetrix Mouse Gene 1.0 ST Array 10500541 - -
          Affymetrix Mouse Gene 1.0 ST Array 10500546 - -
          Affymetrix Mouse Gene 1.0 ST Array 10500535 - -
          Affymetrix Mouse Gene 1.0 ST Array 10500538 - -
          Affymetrix Mouse Gene 1.0 ST Array 10500536 - -
          Affymetrix Mouse Gene 1.0 ST Array 10500542 - -
          Affymetrix HT MG-430 PM Array Plate 1420531_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10500534 - -
          Affymetrix Mouse Gene 1.1 ST Array 10500545 - -
          Affymetrix Mouse Gene 1.1 ST Array 10500539 - -
          Mouse Genome 430 2.0 Array 1420531_PM_at - -
          Mouse Genome 430 2.0 Array 1420531_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15496_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15496_at - -
          Affymetrix Mouse Clariom S Array PSR0300019982.mm.1 NM_008295.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019982.mm.1 NM_008295.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019982.mm.1 TC0300002554.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300019983.mm.1 NM_008295.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019983.mm.1 NM_008295.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300019983.mm.1 TC0300002554.mm.1 MTA TC
          Mouse Expression Set 430 1420531_PM_at - -
          Mouse Expression Set 430 1420531_at - -
          Murine Genome U74v2 Set 94795_PM_at - -
          Murine Genome U74v2 Set 94795_at - -
          Mouse Genome 430A 2.0 Array 1420531_PM_at - -
          Mouse Genome 430A 2.0 Array 1420531_at - -
          Affymetrix HT Mouse Genome 430A Array 1420531_at - -
          Affymetrix HT Mouse Genome 430A Array 1420531_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 522680 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3290112 - -
          Illumina mouse-6 v1.0 expression beadchip 3290112 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003290112 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1234449 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234449 - -
          mouse-6 v1.1 (Illumina) scl21778.5_425-S - -
          Illumina Sentrix mouse-6.v1 bead array 3290112 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21778.5_425-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3290112 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1234449 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234449 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline