Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00657889_mH
          See other CHIL3 GE Assays ›
          Gene Symbol:
          Chil3
          Gene Name:
          chitinase-like 3
          Gene Aliases:
          AI505981, Chi3l3, ECF-L, Ym1
          Chromosome Location:
          Chr.3: 106147554 - 106167564 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00657889_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chil3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009892.2 NP_034022.2 524 90
          GenBank mRNA AK137503.1 - 501 90
          AK154420.1 - 499 90
          BC061154.1 - 499 30298306 90
          D87757.2 - 5 - 6 481 90
          M94584.2 - 493 90
          U56900.1 - 1 - 2 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002684.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chil3

          Panther Classification:

          Molecular Function -

          glycosidase

          Gene Ontology Categories:

          Function(s) Process(es)

          polysaccharide catabolic process
          carbohydrate metabolic process
          chitin catabolic process
          inflammatory response
          metabolic process
          hydrolase activity, hydrolyzing O-glycosyl compounds
          beta-N-acetylhexosaminidase activity
          chitinase activity
          chitin binding
          hydrolase activity
          hydrolase activity, acting on glycosyl bonds
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P167292 D87757.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P167292 M94584.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P167292 AK137503.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P167292 AK154420.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P167292 BC061154.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P167292 D87757.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P167292 M94584.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P167292 AK137503.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P167292 AK154420.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P167292 BC061154.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 122222 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 108335 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 685924 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 5124 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 403616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 316822 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 608218 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 178373 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 611550 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 140040 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 792 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116112 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P167292 D87757.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P167292 M94584.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P167292 AK137503.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P167292 AK154420.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P167292 BC061154.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P167292 D87757.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P167292 M94584.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P167292 AK137503.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P167292 AK154420.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P167292 BC061154.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P167292 D87757.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P167292 M94584.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P167292 AK137503.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P167292 AK154420.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P167292 BC061154.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P167292 D87757.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P167292 M94584.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P167292 AK137503.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P167292 AK154420.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P167292 BC061154.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22400950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22400968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22400965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22400928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22400966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22400938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22400949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22400926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22400954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22400956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22400955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22400946 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05020133 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116112 - -
          Whole Mouse Genome G4122A A_51_P167292 D87757.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P167292 M94584.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P167292 AK137503.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P167292 AK154420.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P167292 BC061154.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P167292 D87757.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P167292 M94584.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P167292 AK137503.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P167292 AK154420.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P167292 BC061154.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104820 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116112 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02974031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22400964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400908 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02903183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400904 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02965887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03100277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03100266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02974051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02965901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02903185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02769938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02965899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02974033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22400887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02974025 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10501025 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501023 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501024 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501021 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501022 - -
          Affymetrix HT MG-430 PM Array Plate 1419764_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10501020 - -
          Mouse Genome 430 2.0 Array 1419764_PM_at - -
          Mouse Genome 430 2.0 Array 1425451_s_at - -
          Mouse Genome 430 2.0 Array 1419764_at - -
          Mouse Genome 430 2.0 Array 1425451_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12655_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12655_at - -
          Affymetrix Mouse Clariom S Array PSR0300020899.mm.1 NM_009892.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020899.mm.1 TC0300002684.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300020902.mm.1 NM_009892.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020902.mm.1 TC0300002684.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300020905.mm.1 NM_009892.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020905.mm.1 TC0300002684.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300020901.mm.1 NM_009892.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020901.mm.1 TC0300002684.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300020903.mm.1 NM_009892.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020903.mm.1 TC0300002684.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300020904.mm.1 NM_009892.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020904.mm.1 TC0300002684.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300020908.mm.1 NM_009892.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020908.mm.1 TC0300002684.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300020910.mm.1 NM_009892.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020910.mm.1 TC0300002684.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300020907.mm.1 NM_009892.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300020907.mm.1 TC0300002684.mm.1 MTA TC
          Mouse Expression Set 430 1425451_s_at - -
          Mouse Expression Set 430 1419764_at - -
          Mouse Expression Set 430 1425451_PM_s_at - -
          Mouse Expression Set 430 1419764_PM_at - -
          Murine Genome U74v2 Set 92694_PM_at - -
          Murine Genome U74v2 Set 92694_at - -
          Mouse Genome 430A 2.0 Array 1425451_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1419764_at - -
          Mouse Genome 430A 2.0 Array 1425451_s_at - -
          Mouse Genome 430A 2.0 Array 1419764_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419764_at - -
          Affymetrix HT Mouse Genome 430A Array 1419764_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 324510 - -
          ABI Mouse Genome Survey Microarray 754283 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6180176 - -
          Illumina mouse-6 v1.0 expression beadchip 6180176 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006180176 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2771793 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2771793 - -
          mouse-6 v1.1 (Illumina) scl012655.1_87-S - -
          Illumina Sentrix mouse-6.v1 bead array 6180176 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl012655.1_87-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6180176 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2712986 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3043036 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3117876 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2712986 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3043036 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3117876 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline