Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00658443_m1
          See other TMEM220 GE Assays ›
          Gene Symbol:
          Tmem220
          Gene Name:
          transmembrane protein 220
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 67025154 - 67035312 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00658443_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem220
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001291042.1 NP_001277971.1 5 - 6 393 153
          NM_001291043.1 NP_001277972.1 5 - 6 389 149
          NM_177392.3 NP_796366.1 6 - 7 465 153
          GenBank mRNA AK043094.1 - 6 - 7 466 153
          AK078505.1 - 5 - 6 392 153
          BC110628.1 - 5 - 6 393 40052084 153
          BC110629.1 - 5 - 6 389 40052085 149

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000848.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem220

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2031133 AK078505.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960688 AK043094.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960688 AK078505.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960688 NM_177392.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 523395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 130295 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 863109 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 315467 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 840185 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 84799 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137957 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121435 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960688 AK043094.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960688 AK078505.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960688 NM_177392.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880469 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27880512 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137957 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121435 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07378777 - -
          Whole Mouse Genome G4122A A_52_P661029 BC110628.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P661029 BC110629.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P661029 AK043094.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P661029 AK078505.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P661029 NM_177392.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101345 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137957 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121435 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09018920 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09667698 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09385766 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09385768 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09385764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27880552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09543487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09094159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09126449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09552039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09018907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09018914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09343285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09676195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09220639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09094157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09696556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09676198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09018929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09543483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09667698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09201873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09543484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27880491 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10377003 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377006 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377004 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377005 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377008 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377002 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377007 - -
          Affymetrix Mouse Gene 1.1 ST Array 10377001 - -
          Affymetrix Mouse Clariom S Array PSR1100006767.mm.1 NM_177392.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006767.mm.1 TC1100000848.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100006768.mm.1 NM_177392.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006768.mm.1 TC1100000848.mm.1 MTA TC
          Murine Genome U74v2 Set 168473_PM_at - -
          Murine Genome U74v2 Set 168473_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 330871 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2060440 - -
          Illumina mouse-6 v1.0 expression beadchip 2060440 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002060440 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228417 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228417 - -
          mouse-6 v1.1 (Illumina) scl41419.7.1_12-S - -
          Illumina Sentrix mouse-6.v1 bead array 2060440 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41419.7.1_12-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2060440 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228417 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228417 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline