Search
Search
| Gene Symbol: Cd74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042605.1 | NP_001036070.1 | 1 - 2 | 159 | 118 | |
| NM_010545.3 | NP_034675.1 | 1 - 2 | 159 | 118 | ||
| GenBank mRNA | AK002232.1 | - | 1 - 2 | 159 | 118 | |
| AK020607.1 | - | 1 - 2 | 160 | 118 | ||
| AK154096.1 | - | 1 - 2 | 190 | 118 | ||
| BC003476.1 | - | 1 - 2 | 159 | 2650401 | 118 | |
| BC096435.1 | - | 1 - 2 | 170 | 6530802 | 118 | |
| X07129.1 | - | 1 - 2 | 152 | 118 | ||
Set Membership: |
|