Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00658599_g1
          See other KLK1B4 GE Assays ›
          Gene Symbol:
          Klk1b4
          Gene Name:
          kallikrein 1-related pepidase b4
          Gene Aliases:
          Ngfa, mGk-4
          Chromosome Location:
          Chr.7: 44207435 - 44211754 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00658599_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klk1b4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010915.3 NP_035045.2 636 135
          GenBank mRNA BC034518.1 - 636 4911885 135
          M11434.1 - 632 135

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klk1b4

          Panther Classification:

          Molecular Function -

          protease serine protease protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          small GTPase mediated signal transduction
          positive regulation of phosphatidylinositol 3-kinase activity
          serine-type endopeptidase activity
          receptor signaling protein activity
          growth factor activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295034 NM_010915.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295034 BC034518.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295034 M11434.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295034 NM_010915.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295034 BC034518.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P295034 M11434.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 289506 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 109964 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103129 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295034 NM_010915.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295034 BC034518.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295034 M11434.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295034 NM_010915.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295034 BC034518.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P295034 M11434.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295034 NM_010915.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295034 BC034518.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295034 M11434.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295034 NM_010915.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295034 BC034518.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P295034 M11434.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25086092 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P103129 - -
          Whole Mouse Genome G4122A A_51_P295034 NM_010915.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P295034 BC034518.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P295034 M11434.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P295034 NM_010915.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P295034 BC034518.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P295034 M11434.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P103129 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05930434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25086043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06482869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06409252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06546985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06398676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25086107 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552548 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552551 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552617 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552552 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552616 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552614 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552615 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552550 - -
          Affymetrix HT MG-430 PM Array Plate 1421372_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1450222_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10552546 - -
          Affymetrix Mouse Gene 1.1 ST Array 10552613 - -
          Mouse Genome 430 2.0 Array 1450222_PM_x_at - -
          Mouse Genome 430 2.0 Array 1421372_at - -
          Mouse Genome 430 2.0 Array 1450222_x_at - -
          Mouse Genome 430 2.0 Array 1421372_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18048_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18048_PM_at - -
          Mouse Expression Set 430 1450222_x_at - -
          Mouse Expression Set 430 1450222_PM_x_at - -
          Mouse Expression Set 430 1421372_at - -
          Mouse Expression Set 430 1421372_PM_at - -
          Murine Genome U74v2 Set 94716_f_at - -
          Murine Genome U74v2 Set 94773_at - -
          Murine Genome U74v2 Set 94773_PM_at - -
          Murine Genome U74v2 Set 94716_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1421372_at - -
          Mouse Genome 430A 2.0 Array 1421372_PM_at - -
          Mouse Genome 430A 2.0 Array 1450222_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1450222_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1421372_at - -
          Affymetrix HT Mouse Genome 430A Array 1450222_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1421372_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450222_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 785156 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 630348 - -
          MouseWG-6 v2 Expression BeadChip 5900605 - -
          Illumina mouse-6 v1.0 expression beadchip 100130301 - -
          Illumina mouse-6 v1.0 expression beadchip 630348 - -
          Illumina mouse-6 v1.0 expression beadchip 5900605 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005900605 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000630348 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2697256 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2697256 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253983 - -
          mouse-6 v1.1 (Illumina) GI_8659571-S - -
          mouse-6 v1.1 (Illumina) scl32723.5.1_67-S - -
          mouse-6 v1.1 (Illumina) scl018050.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 630348 - -
          Illumina Sentrix mouse-6.v1 bead array 5900605 - -
          Illumina Sentrix mouse-6.v1 bead array 100130301 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32723.5.1_67-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl018050.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5900605 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100130301 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 630348 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2697256 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1238736 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2697256 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238736 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1253983 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl018050.1_7-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline