Search
Search

Trp53bp1| Gene Symbol: Trp53bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290830.1 | NP_001277759.1 | 20 - 21 | 4633 | 66 | |
| NM_013735.4 | NP_038763.3 | 21 - 22 | 4783 | 66 | ||
| GenBank mRNA | AJ414734.1 | - | 20 - 21 | 4626 | 66 | |
| AK029832.1 | - | 1180 | 66 | |||
| AK036324.1 | - | 2 - 3 | 588 | 66 | ||
| AK132323.1 | - | 3 - 4 | 816 | 66 | ||
| BC035206.1 | - | 9 - 10 | 1995 | 3979746 | 66 | |
| BC079906.1 | - | 20 - 21 | 4567 | 30639375 | 66 | |
Set Membership: |
|