Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00659817_m1
          See other NUMA1 GE Assays ›
          Gene Symbol:
          Numa1
          Gene Name:
          nuclear mitotic apparatus protein 1
          Gene Aliases:
          6720401E04Rik, AA764025, AL022610, AU014979
          Chromosome Location:
          Chr.7: 101934002 - 102014959 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00659817_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Numa1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133947.3 NP_598708.3 12 - 13 1268 118
          GenBank mRNA AK032693.1 - 12 - 13 1317 118
          AK050670.1 - 12 - 13 1350 118
          AK168399.1 - 12 - 13 1286 118
          BC049791.1 - 12 - 13 1268 6505037 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001444.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Numa1

          Panther Classification:

          Molecular Function -

          cytoskeletal protein microtubule or microtubule-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of mitotic spindle orientation
          lung epithelial cell differentiation
          microtubule binding
          tubulin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338461 BC006631.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338461 BC049791.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338461 BC004667.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338461 AK144141.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338461 AK168399.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338461 BC006631.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338461 BC049791.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338461 BC004667.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338461 AK144141.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P338461 AK168399.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 708377 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 576381 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633956 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 683903 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 608926 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704743 AF227731.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704743 AF249296.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704743 AK004822.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704743 AK004899.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704743 BC029718.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704743 BC037086.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704743 NM_022722.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 769012 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 609164 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 121656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 603160 NM_001077425.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 24779 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 592947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 890881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 383825 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 65759 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848293 AY173049.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848293 NM_172916.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 831870 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193874 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 350148 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865599 AK078940.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865599 AK137529.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865599 AK153136.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865599 BC053105.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865599 NM_199446.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881283 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 253454 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177538 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 683737 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33984 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35261 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33828 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 27131 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 36249 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129713 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143169 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118104 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338461 BC006631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338461 BC049791.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338461 BC004667.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338461 AK144141.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338461 AK168399.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338461 BC006631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338461 BC049791.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338461 BC004667.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338461 AK144141.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P338461 AK168399.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P298473 BC049791.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P298473 AK168399.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P298473 AK050670.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P298473 NM_133947.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338461 BC006631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338461 BC049791.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338461 BC004667.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338461 AK144141.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338461 AK168399.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338461 BC006631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338461 BC049791.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338461 BC004667.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338461 AK144141.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P338461 AK168399.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111763 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06605777 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06605778 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06605773 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06430215 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05914868 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06430222 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06605768 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06605772 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06430220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25376293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06044549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06642954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06223097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06460579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06645505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06547966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05942946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06390391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05942943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06547973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06427249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25376260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06577219 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555599 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555612 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555606 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555605 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555619 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555598 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555603 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555611 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555602 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555618 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555609 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555615 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555613 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555593 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555601 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555592 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555614 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555594 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555604 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555591 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555616 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555617 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555600 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555610 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555596 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555597 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555607 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555595 - -
          Affymetrix HT MG-430 PM Array Plate 1423974_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1441090_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423975_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10555590 - -
          Mouse Genome 430 2.0 Array 1441090_PM_at - -
          Mouse Genome 430 2.0 Array 1423974_at - -
          Mouse Genome 430 2.0 Array 1423974_PM_at - -
          Mouse Genome 430 2.0 Array 1441090_at - -
          Mouse Genome 430 2.0 Array 1423975_PM_s_at - -
          Mouse Genome 430 2.0 Array 1423975_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 101706_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 101706_at - -
          Affymetrix Mouse Clariom S Array PSR0700012450.mm.1 TC0700001444.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012450.mm.1 NM_133947.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012450.mm.1 NM_133947.3 Refseq
          Mouse Expression Set 430 1423975_PM_s_at - -
          Mouse Expression Set 430 1423974_PM_at - -
          Mouse Expression Set 430 1423974_at - -
          Mouse Expression Set 430 1423975_s_at - -
          Mouse Expression Set 430 1441090_at - -
          Mouse Expression Set 430 1441090_PM_at - -
          Murine Genome U74v2 Set 94976_PM_at - -
          Murine Genome U74v2 Set 94976_at - -
          Mouse Genome 430A 2.0 Array 1423974_PM_at - -
          Mouse Genome 430A 2.0 Array 1423975_s_at - -
          Mouse Genome 430A 2.0 Array 1423975_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1423974_at - -
          Affymetrix HT Mouse Genome 430A Array 1423974_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423974_at - -
          Affymetrix HT Mouse Genome 430A Array 1423975_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1423975_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1441090_at - -
          Affymetrix HT Mouse Genome 430B Array 1441090_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 918718 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6520576 - -
          Illumina mouse-6 v1.0 expression beadchip 6520576 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006520576 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2735625 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2735625 - -
          mouse-6 v1.1 (Illumina) scl32286.29.1_5-S - -
          Illumina Sentrix mouse-6.v1 bead array 6520576 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32286.29.1_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6520576 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2735625 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2735625 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl32286.29.1_5-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline