Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00659987_m1
          See other TMEM120A GE Assays ›
          Gene Symbol:
          Tmem120a
          Gene Name:
          transmembrane protein 120A
          Gene Aliases:
          2010310D06Rik, Tmpit
          Chromosome Location:
          Chr.5: 135735490 - 135744172 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00659987_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem120a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172541.2 NP_766129.1 3 - 4 365 136
          GenBank mRNA AK028119.1 - 3 - 4 366 136
          BC046757.1 - 3 - 4 325 5693443 136

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003404.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem120a

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24117668 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24117666 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24117665 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24117669 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333349 AK028119.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333349 AK050367.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333349 AK209822.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333349 AK211918.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333349 NM_172541.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333349 AK028119.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333349 AK050367.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333349 AK209822.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333349 AK211918.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P333349 NM_172541.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 137078 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397247 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434245 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 369180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 782238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 134441 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129541 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333349 AK028119.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333349 AK050367.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333349 AK209822.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333349 AK211918.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333349 NM_172541.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333349 AK028119.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333349 AK050367.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333349 AK209822.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333349 AK211918.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P333349 NM_172541.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24117670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24117650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24117658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24117661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24117653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24117656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24117681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24117660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24117673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24117663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24117666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24117665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24117678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24117652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24117675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24117671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24117685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24117668 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P129541 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05756313 - -
          Whole Mouse Genome G4122A A_51_P333349 AK028119.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333349 AK050367.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333349 AK209822.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333349 AK211918.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333349 NM_172541.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333349 AK028119.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333349 AK050367.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333349 AK209822.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333349 AK211918.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333349 NM_172541.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134180 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138441 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P129541 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04766942 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04419398 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24117669 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24117666 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24117668 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24117665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24117642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24117665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24117645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04766945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04461997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24117668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24117642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24117678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04461995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04492791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24117656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24117638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04533837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24117663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24117681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24117672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24117676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24117671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04321874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24117675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04733555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24117674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24117650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04485782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24117660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04492800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24117661 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10534518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534513 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534510 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534517 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534507 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534508 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534505 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534509 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534512 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534506 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534514 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534515 - -
          Affymetrix HT MG-430 PM Array Plate 1435031_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10534504 - -
          Mouse Genome 430 2.0 Array 1435031_PM_at - -
          Mouse Genome 430 2.0 Array 1435031_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 215210_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 215210_at - -
          Affymetrix Mouse Clariom S Array PSR0500030758.mm.1 NM_172541.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030758.mm.1 NM_172541.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030758.mm.1 TC0500003404.mm.1 MTA TC
          Mouse Expression Set 430 1435031_PM_at - -
          Mouse Expression Set 430 1435031_at - -
          Affymetrix HT Mouse Genome 430B Array 1435031_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435031_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 381640 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3830538 - -
          Illumina mouse-6 v1.0 expression beadchip 3830538 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003830538 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2509273 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2509273 - -
          mouse-6 v1.1 (Illumina) scl25921.12.1_22-S - -
          Illumina Sentrix mouse-6.v1 bead array 3830538 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25921.12.1_22-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3830538 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2509273 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2509273 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl25921.12.1_22-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline