Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00660410_m1
          See other ACBD4 GE Assays ›
          Gene Symbol:
          Acbd4
          Gene Name:
          acyl-Coenzyme A binding domain containing 4
          Gene Aliases:
          2010009P05Rik, 2010015A21Rik, AI849317
          Chromosome Location:
          Chr.11: 103101688 - 103112199 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00660410_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acbd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025988.2 NP_080264.1 9 - 10 1424 97
          GenBank mRNA AK008207.1 - 3 - 4 484 97
          AK008243.1 - 7 - 8 973 97
          AK079604.1 - 8 - 9 1293 97
          AK165381.1 - 3 - 4 561 97
          AK218656.1 - 1 - 2 53 97
          BC048371.1 - 8 - 9 1338 6396076 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001705.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acbd4

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083324 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292757 AK165381.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292757 BC048371.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292757 AK079604.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292757 AK148438.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292757 NM_025988.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292757 AK165381.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292757 BC048371.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292757 AK079604.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292757 AK148438.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P292757 NM_025988.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2139611 AK165381.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2139611 BC048371.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2139611 AK079604.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2139611 AK148438.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2139611 NM_025988.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 40947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 552634 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 791548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 954803 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727871 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 458960 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360128 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 814092 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 349615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 78299 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P180414 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165322 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292757 AK165381.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292757 BC048371.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292757 AK079604.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292757 AK148438.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292757 NM_025988.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292757 AK165381.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292757 BC048371.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292757 AK079604.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292757 AK148438.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P292757 NM_025988.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2139611 AK165381.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2139611 BC048371.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2139611 AK079604.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2139611 AK148438.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2139611 NM_025988.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292757 AK165381.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292757 BC048371.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292757 AK079604.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292757 AK148438.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292757 NM_025988.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292757 AK165381.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292757 BC048371.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292757 AK079604.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292757 AK148438.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P292757 NM_025988.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28083358 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P180414 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07471292 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165322 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07471274 - -
          Whole Mouse Genome G4122A A_51_P292757 AK165381.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292757 BC048371.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292757 AK079604.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292757 AK148438.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292757 NM_025988.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292757 AK165381.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292757 BC048371.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292757 AK079604.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292757 AK148438.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P292757 NM_025988.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129898 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P180414 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165322 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28083324 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10381686 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381687 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381694 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381693 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381685 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381689 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381692 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381684 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381691 - -
          Affymetrix HT MG-430 PM Array Plate 1428271_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1449626_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10381683 - -
          Mouse Genome 430 2.0 Array 1449626_PM_s_at - -
          Mouse Genome 430 2.0 Array 1449626_s_at - -
          Mouse Genome 430 2.0 Array 1428271_at - -
          Mouse Genome 430 2.0 Array 1428271_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67131_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67131_at - -
          Affymetrix Mouse Clariom S Array PSR1100016117.mm.1 NM_025988.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016117.mm.1 NM_025988.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016117.mm.1 TC1100001705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016109.mm.1 NM_025988.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016109.mm.1 NM_025988.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016109.mm.1 TC1100001705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016114.mm.1 NM_025988.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016114.mm.1 NM_025988.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016114.mm.1 TC1100001705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016115.mm.1 NM_025988.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016115.mm.1 NM_025988.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016115.mm.1 TC1100001705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016113.mm.1 NM_025988.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016113.mm.1 NM_025988.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016113.mm.1 TC1100001705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016116.mm.1 NM_025988.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016116.mm.1 NM_025988.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016116.mm.1 TC1100001705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016118.mm.1 NM_025988.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016118.mm.1 NM_025988.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016118.mm.1 TC1100001705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016111.mm.1 NM_025988.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016111.mm.1 NM_025988.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016111.mm.1 TC1100001705.mm.1 MTA TC
          Mouse Expression Set 430 1428271_PM_at - -
          Mouse Expression Set 430 1428271_at - -
          Affymetrix HT Mouse Genome 430B Array 1428271_at - -
          Affymetrix HT Mouse Genome 430B Array 1428271_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1449626_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1449626_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6980079 - -
          Illumina mouse-6 v1.0 expression beadchip 6980079 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006980079 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2689540 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2689540 - -
          mouse-6 v1.1 (Illumina) scl40849.10_252-S - -
          Illumina Sentrix mouse-6.v1 bead array 6980079 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40849.10_252-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6980079 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3150296 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2689540 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3071515 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3150296 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3071515 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2689540 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40849.10_252-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0