Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00660631_m1
          See other TAF11 GE Assays ›
          Gene Symbol:
          Taf11
          Gene Name:
          TAF11 RNA polymerase II, TATA box binding protein (TBP)-associated factor
          Gene Aliases:
          1110038O14Rik, 28kDa, AI115401, AL024207, AW552398, PRO214, TAF2I, TAFII28
          Chromosome Location:
          Chr.17: 27901128 - 27909242 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00660631_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Taf11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026836.2 NP_081112.1 1 - 2 220 75
          GenBank mRNA AK050689.1 - 1 - 2 221 75
          AK133851.1 - 2 - 3 1204 75
          BC005603.1 - 1 - 2 199 3483315 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001747.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Taf11

          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription initiation from RNA polymerase II promoter
          positive regulation by host of viral transcription
          RNA polymerase II transcriptional preinitiation complex assembly
          regulation of nucleic acid-templated transcription
          DNA binding
          transcription coactivator activity
          transcription factor binding
          vitamin D receptor binding
          thyroid hormone receptor binding
          protein heterodimerization activity
          protein N-terminus binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31119189 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31119191 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31119192 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31119182 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31119183 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31119180 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31119190 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31119181 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092073 NM_026836.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092073 AK050689.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092073 AK133851.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 267295 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375507 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 668119 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679401 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669553 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510687 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 12199 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 24853 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169388 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092073 NM_026836.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092073 AK050689.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092073 AK133851.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P410127 BC005603.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P410127 NM_026836.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P410127 AK004168.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P410127 AK050689.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P410127 AK133851.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P230628 BC005603.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P230628 NM_026836.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P230628 AK004168.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P230628 AK050689.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P230628 AK133851.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31119171 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169388 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08749285 - -
          Whole Mouse Genome G4122A A_51_P410127 BC005603.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P410127 NM_026836.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P410127 AK004168.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P410127 AK050689.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P410127 AK133851.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P230628 BC005603.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P230628 NM_026836.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P230628 AK004168.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P230628 AK050689.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P230628 AK133851.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132145 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169388 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13203896 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13305204 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13305203 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13305195 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13305201 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13305206 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31119180 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31119183 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31119192 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31119181 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31119182 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31119189 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31119190 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31119191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31119203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13537971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13305206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13305203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31119173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13429117 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10449400 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449395 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449397 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449399 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449396 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449398 - -
          Affymetrix HT MG-430 PM Array Plate 1451995_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10449394 - -
          Mouse Genome 430 2.0 Array 1446161_at - -
          Mouse Genome 430 2.0 Array 1446161_PM_at - -
          Mouse Genome 430 2.0 Array 1451995_PM_at - -
          Mouse Genome 430 2.0 Array 1451995_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68776_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68776_at - -
          Affymetrix Mouse Clariom S Array PSR1700016088.mm.1 NM_026836.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016088.mm.1 NM_026836.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016088.mm.1 TC1700001747.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700016090.mm.1 NM_026836.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016090.mm.1 NM_026836.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016090.mm.1 TC1700001747.mm.1 MTA TC
          Mouse Expression Set 430 1451995_at - -
          Mouse Expression Set 430 1451995_PM_at - -
          Murine Genome U74v2 Set 96691_PM_at - -
          Murine Genome U74v2 Set 96691_at - -
          Murine Genome U74v2 Set 170787_r_at - -
          Murine Genome U74v2 Set 170704_PM_i_at - -
          Murine Genome U74v2 Set 170787_PM_r_at - -
          Murine Genome U74v2 Set 170704_i_at - -
          Mouse Genome 430A 2.0 Array 1451995_PM_at - -
          Mouse Genome 430A 2.0 Array 1451995_at - -
          Affymetrix HT Mouse Genome 430A Array 1451995_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451995_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 761588 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5720538 - -
          Illumina mouse-6 v1.0 expression beadchip 5720538 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005720538 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223605 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223605 - -
          mouse-6 v1.1 (Illumina) scl068776.1_229-S - -
          Illumina Sentrix mouse-6.v1 bead array 5720538 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl068776.1_229-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5720538 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2946281 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2946281 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl068776.1_229-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline