| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24045560 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24045561 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24045559 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2088490 |
BC029047.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
639726 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
202370 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
505282 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
283746 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
130932 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
659399 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
203921 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855922 |
AB041569.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855922 |
AK043957.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855922 |
AK140685.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855922 |
AK141493.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855922 |
BC043329.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855922 |
EF537862.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855922 |
EF537863.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
855922 |
NM_021427.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
325788 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
928465 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
543583 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
83610 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
517465 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
369341 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
245323 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
474436 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
170583 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
496570 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
211181 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
934757 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
666772 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
626551 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
517148 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
60834 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
175425 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
171005 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P167007 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P105387 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2088490 |
BC029047.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P187507 |
BC027825.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P187507 |
BC029047.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P187507 |
AK050332.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P187507 |
NM_028037.4 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P187507 |
AK010568.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24045785 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24045778 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24045767 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24045780 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24045777 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24045781 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24045756 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P105387 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P167007 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01205927 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P05722458 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01205974 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P187507 |
BC027825.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P187507 |
BC029047.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P187507 |
AK050332.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P187507 |
NM_028037.4 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P187507 |
AK010568.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P123518 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P114502 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P105387 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P167007 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04696925 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P04696926 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24045561 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24045560 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24045559 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045684 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045743 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045698 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045524 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045714 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045696 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045697 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045785 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045621 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045721 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045528 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045732 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045649 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045599 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045711 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045579 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045739 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045564 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045554 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045557 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045692 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045668 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045665 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045643 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045662 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045778 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045623 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045527 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045616 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045713 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045651 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045691 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045747 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045719 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045704 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045716 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045780 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045745 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045612 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045575 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045558 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045726 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045727 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045648 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045667 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045709 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045636 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045638 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045694 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045715 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045611 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045655 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045540 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045751 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045670 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045572 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045688 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045707 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045577 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045725 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045653 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045640 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045737 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045702 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045573 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045729 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045676 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045562 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045738 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045559 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045749 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045685 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045723 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045681 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045619 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045589 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045706 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045724 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045652 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045701 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045728 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045555 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045678 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045710 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24045699 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045660 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045645 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04197516 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04518037 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045621 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04606812 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045623 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045570 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045651 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045613 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04850666 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045785 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045524 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04470870 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04227946 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045676 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045745 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04278965 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045572 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045684 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045589 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04545517 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045653 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045667 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045644 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04828647 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04197512 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04352658 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04961386 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045527 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045656 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045606 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045573 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045685 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045552 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04343602 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045523 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04333661 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045781 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045530 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045674 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045743 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045638 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04828652 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045555 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04976649 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04961389 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045520 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045554 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045773 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04227948 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04197510 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04227947 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045636 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045680 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045648 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04523712 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04976101 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045681 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04518047 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045778 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04949398 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04752130 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045528 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045619 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04550666 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045521 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04696926 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04545518 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045591 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04390938 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045767 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045639 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04473607 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045588 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04512000 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045579 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045626 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045780 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045753 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045611 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04713400 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045670 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045657 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045748 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04579754 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045649 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04159255 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045609 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045612 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045659 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045673 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04545513 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04961388 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045749 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P04752129 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045616 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P24045686 |
- |
- |