Search
Search

Usp20| Gene Symbol: Usp20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028846.5 | NP_083122.1 | 22 - 23 | 2654 | 62 | |
| GenBank mRNA | AF449715.1 | - | 22 - 23 | 2605 | 62 | |
| AK006800.1 | - | 5 - 6 | 588 | 62 | ||
| AK029890.1 | - | 2550 | 62 | |||
| AK047242.1 | - | 22 - 23 | 2573 | 62 | ||
| AK054279.1 | - | 22 - 23 | 2597 | 62 | ||
| AK087412.1 | - | 22 - 23 | 2566 | 62 | ||
| AK163663.1 | - | 22 - 23 | 2564 | 62 | ||
| AK173083.1 | - | 22 - 23 | 2556 | 62 | ||
| AK215664.1 | - | 1 - 2 | 104 | 62 | ||
| BC019735.1 | - | 8 - 9 | 854 | 62 | ||
| BC037199.1 | - | 16 - 17 | 2038 | 62 | ||
| BC079674.1 | - | 2543 | 30531994 | 62 | ||
Set Membership: |
|