Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00661433_s1
          See other PRPS1L1 GE Assays ›
          Gene Symbol:
          Prps1l1
          Gene Name:
          phosphoribosyl pyrophosphate synthetase 1-like 1
          Gene Aliases:
          1700011K15Rik
          Chromosome Location:
          Chr.12: 34984761 - 34986436 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00661433_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prps1l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029294.2 NP_083570.1 1 1171 124
          GenBank mRNA AK077203.1 - 1 1172 124

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prps1l1

          Panther Classification:

          Molecular Function -

          nucleotide kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443314 AK077203.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443314 NM_029294.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443314 AK077203.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443314 NM_029294.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151238 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443314 AK077203.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443314 NM_029294.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443314 AK077203.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443314 NM_029294.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28350675 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151238 - -
          Whole Mouse Genome G4122A A_51_P443314 AK077203.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P443314 NM_029294.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P443314 AK077203.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P443314 NM_029294.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107658 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151238 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10117047 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09871163 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09871142 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10009833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28350709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09929252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10425246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09871149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09871564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09929256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10108666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09918747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10168324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09871161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09952375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10032777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09871142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10117045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28350658 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10395326 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395325 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395327 - -
          Affymetrix HT MG-430 PM Array Plate 1453203_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10395324 - -
          Mouse Genome 430 2.0 Array 1453203_at - -
          Mouse Genome 430 2.0 Array 1453203_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75456_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75456_at - -
          Affymetrix Mouse Clariom S Array PSR1200002296.mm.1 NM_029294.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200002296.mm.1 NM_029294.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200002296.mm.1 TC1200000301.mm.1 MTA TC
          Mouse Expression Set 430 1453203_PM_at - -
          Mouse Expression Set 430 1453203_at - -
          Murine Genome U74v2 Set 166251_f_at - -
          Murine Genome U74v2 Set 168800_PM_at - -
          Murine Genome U74v2 Set 168800_at - -
          Murine Genome U74v2 Set 166251_PM_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1453203_at - -
          Affymetrix HT Mouse Genome 430B Array 1453203_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 490520 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1850411 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001850411 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221385 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221385 - -
          mouse-6 v1.1 (Illumina) scl43289.1.46_42-S - -
          Illumina Sentrix mouse-6.v1 bead array 1850411 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl43289.1.46_42-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1850411 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2925214 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1221385 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221385 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2925214 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline