Search
Search

Golga1| Gene Symbol: Golga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290649.1 | NP_001277578.1 | 13 - 14 | 1579 | 74 | |
| NM_029793.2 | NP_084069.1 | 14 - 15 | 1654 | 74 | ||
| NR_110968.1 | 14 - 15 | 1654 | 74 | |||
| GenBank mRNA | AK028516.1 | - | 14 - 15 | 1574 | 74 | |
| AK044987.1 | - | 13 - 14 | 2064 | 74 | ||
| BC037037.1 | - | 9 - 10 | 990 | 74 | ||
| BC043090.1 | - | 14 - 15 | 1519 | 6418353 | 74 | |
| BC053068.1 | - | 13 - 14 | 1507 | 6405595 | 74 | |
| BC080733.1 | - | 13 - 14 | 1579 | 30110235 | 74 | |
Set Membership: |
|