Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00661630_m1
          See other CLCA3A2 GE Assays ›
          Gene Symbol:
          Clca3a2
          Gene Name:
          chloride channel calcium activated 3A2
          Gene Aliases:
          Clca2
          Chromosome Location:
          Chr.3: 144796559 - 144819629 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00661630_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clca3a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030601.3 NP_085104.1 4 - 5 572 92
          GenBank mRNA AF108501.1 - 4 - 5 563 92
          AF115852.1 - 4 - 5 563 92
          AK042314.1 - 4 - 5 565 92
          AK132498.1 - 4 - 5 572 92
          AK161052.1 - 4 - 5 565 92
          BC008147.1 - 4 - 5 566 3583804 92
          BC039166.1 - 4 - 5 555 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300003252.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Ion Channel 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clca3a2

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          chloride transport
          anoikis
          extrinsic apoptotic signaling pathway in absence of ligand
          chloride transmembrane transport
          intracellular calcium activated chloride channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P265806 BC039166.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P265806 AK161052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P265806 BC008147.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P265806 AK132498.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P265806 AK134192.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P265806 BC039166.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P265806 AK161052.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P265806 BC008147.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P265806 AK132498.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P265806 AK134192.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701399 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 869165 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351065 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 386716 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 801073 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 100004 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167719 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 131383 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119160 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P265806 BC039166.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P265806 AK161052.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P265806 BC008147.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P265806 AK132498.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P265806 AK134192.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P265806 BC039166.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P265806 AK161052.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P265806 BC008147.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P265806 AK132498.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P265806 AK134192.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P265806 BC039166.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P265806 AK161052.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P265806 BC008147.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P265806 AK132498.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P265806 AK134192.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P265806 BC039166.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P265806 AK161052.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P265806 BC008147.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P265806 AK132498.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P265806 AK134192.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605170 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05105201 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00779779 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119160 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05105213 - -
          Whole Mouse Genome G4122A A_51_P265806 BC039166.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P265806 AK161052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P265806 BC008147.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P265806 AK132498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P265806 AK134192.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P265806 BC039166.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P265806 AK161052.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P265806 BC008147.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P265806 AK132498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P265806 AK134192.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119160 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03068898 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02725033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02725035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02424854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02424852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03127535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03127538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605136 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10502567 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502572 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502573 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502566 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502569 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502570 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502574 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502568 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502571 - -
          Affymetrix HT MG-430 PM Array Plate 1419463_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1437578_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10502565 - -
          Mouse Genome 430 2.0 Array 1437578_PM_at - -
          Mouse Genome 430 2.0 Array 1460259_s_at - -
          Mouse Genome 430 2.0 Array 1460259_PM_s_at - -
          Mouse Genome 430 2.0 Array 1437578_at - -
          Mouse Genome 430 2.0 Array 1419463_PM_at - -
          Mouse Genome 430 2.0 Array 1419463_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 80797_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 80797_at - -
          Affymetrix Mouse Clariom S Array PSR0300024187.mm.1 NM_030601.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024187.mm.1 NM_030601.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024187.mm.1 TC0300003252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024189.mm.1 NM_030601.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024189.mm.1 NM_030601.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024189.mm.1 TC0300003252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024197.mm.1 NM_030601.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024197.mm.1 NM_030601.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024197.mm.1 TC0300003252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024203.mm.1 NM_030601.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024203.mm.1 NM_030601.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024203.mm.1 TC0300003252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024190.mm.1 NM_030601.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024190.mm.1 NM_030601.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024190.mm.1 TC0300003252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024192.mm.1 NM_030601.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024192.mm.1 NM_030601.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024192.mm.1 TC0300003252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024196.mm.1 NM_030601.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024196.mm.1 NM_030601.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024196.mm.1 TC0300003252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024188.mm.1 NM_030601.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024188.mm.1 NM_030601.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024188.mm.1 TC0300003252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024195.mm.1 NM_030601.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024195.mm.1 NM_030601.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024195.mm.1 TC0300003252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024200.mm.1 NM_030601.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024200.mm.1 NM_030601.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024200.mm.1 TC0300003252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024191.mm.1 NM_030601.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024191.mm.1 NM_030601.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024191.mm.1 TC0300003252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024193.mm.1 NM_030601.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024193.mm.1 NM_030601.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024193.mm.1 TC0300003252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024194.mm.1 NM_030601.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024194.mm.1 NM_030601.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024194.mm.1 TC0300003252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024199.mm.1 NM_030601.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024199.mm.1 NM_030601.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024199.mm.1 TC0300003252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024201.mm.1 NM_030601.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024201.mm.1 NM_030601.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024201.mm.1 TC0300003252.mm.1 MTA TC
          Mouse Expression Set 430 1419463_PM_at - -
          Mouse Expression Set 430 1460259_s_at - -
          Mouse Expression Set 430 1419463_at - -
          Mouse Expression Set 430 1460259_PM_s_at - -
          Mouse Expression Set 430 1437578_PM_at - -
          Mouse Expression Set 430 1437578_at - -
          Mouse Genome 430A 2.0 Array 1460259_s_at - -
          Mouse Genome 430A 2.0 Array 1419463_PM_at - -
          Mouse Genome 430A 2.0 Array 1419463_at - -
          Mouse Genome 430A 2.0 Array 1460259_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1419463_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419463_at - -
          Affymetrix HT Mouse Genome 430B Array 1437578_at - -
          Affymetrix HT Mouse Genome 430B Array 1437578_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 387241 - -
          ABI Mouse Genome Survey Microarray 750894 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2230519 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002230519 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2761361 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2761361 - -
          mouse-6 v1.1 (Illumina) scl21420.15_12-S - -
          Illumina Sentrix mouse-6.v1 bead array 2230519 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21420.15_12-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2230519 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2761361 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2761361 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline