Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00661737_m1
          See other NXF7 GE Assays ›
          Gene Symbol:
          Nxf7
          Gene Name:
          nuclear RNA export factor 7
          Gene Aliases:
          Nx7, Nxf6, nfx-a1, nxf-a, nxf-a2
          Chromosome Location:
          Chr.X: 135579555 - 135598777 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00661737_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nxf7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_130888.1 NP_570958.1 11 - 12 1030 66
          GenBank mRNA AB178901.1 - 12 - 13 1169 66
          AY260550.1 - 11 - 12 1030 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00003012.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nxf7

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Process(es)

          RNA binding
          mRNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102449 AJ305318.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102449 AB178901.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2102449 AJ305317.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163093 AJ305318.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163093 AY260550.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163093 NM_130888.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163093 AB178901.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2163093 AJ305317.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 373671 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 620019 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 856585 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 520028 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360319 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 257694 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 259214 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 121729 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493153 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 825720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122775 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111562 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141530 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163093 AJ305318.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163093 AY260550.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163093 NM_130888.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163093 AB178901.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2163093 AJ305317.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677474 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677485 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677479 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677492 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677471 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32677509 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09428358 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09428303 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111562 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141530 - -
          Whole Mouse Genome G4122A A_52_P460570 AJ305318.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P460570 AY260550.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P460570 NM_130888.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P460570 AB178901.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P460570 AJ305317.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139136 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141530 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111562 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15215671 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15215672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32677509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14576915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14576917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14593249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14913296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14776702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14804646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14776711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14776631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14776667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14576914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677502 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14776636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14997822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14776682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677440 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14776699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14568149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14776646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32677503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14804644 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10606799 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606810 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606800 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606797 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606814 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606796 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606811 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606803 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606798 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606804 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606809 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606806 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606812 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606813 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606793 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606808 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606794 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606795 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606807 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606801 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606805 - -
          Affymetrix Mouse Gene 1.0 ST Array 10606802 - -
          Affymetrix HT MG-430 PM Array Plate 1427399_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10606792 - -
          Mouse Genome 430 2.0 Array 1427399_a_at - -
          Mouse Genome 430 2.0 Array 1427399_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170722_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170722_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00018846.mm.1 NM_130888.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018846.mm.1 NM_130888.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018846.mm.1 TC0X00003012.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00018847.mm.1 NM_130888.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018847.mm.1 NM_130888.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00018847.mm.1 TC0X00003012.mm.1 MTA TC
          Mouse Expression Set 430 1427399_a_at - -
          Mouse Expression Set 430 1427399_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1427399_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1427399_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427399_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427399_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 474197 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4850025 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004850025 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2629335 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2629335 - -
          mouse-6 v1.1 (Illumina) scl53823.22.1_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 4850025 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53823.22.1_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4850025 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2629335 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2629335 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline