Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00662226_gH
          See other REEP4 GE Assays ›
          Gene Symbol:
          Reep4
          Gene Name:
          receptor accessory protein 4
          Gene Aliases:
          2700029E10Rik
          Chromosome Location:
          Chr.14: 70545218 - 70548935 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00662226_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Reep4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_180588.2 NP_850919.1 7 - 8 1163 64
          GenBank mRNA AK012301.1 - 7 - 8 1060 64
          AY562232.1 - 7 - 8 707 64
          BC033929.1 - 7 - 8 791 1363539 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001000.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Reep4

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear envelope organization
          cell cycle
          mitotic nuclear division
          mitotic nuclear envelope reassembly
          cell division
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29701959 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29701961 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29701957 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29701960 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29701958 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29701956 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P420655 AK012301.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P420655 BC033929.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P420655 NM_180588.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P420655 AK012301.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P420655 BC033929.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P420655 NM_180588.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84521 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130031 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P420655 AK012301.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P420655 BC033929.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P420655 NM_180588.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P420655 AK012301.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P420655 BC033929.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P420655 NM_180588.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29701956 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130031 - -
          Whole Mouse Genome G4122A A_51_P420655 AK012301.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P420655 BC033929.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P420655 NM_180588.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P420655 AK012301.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P420655 BC033929.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P420655 NM_180588.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134101 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130031 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11555259 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11555254 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11732525 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11555252 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11732527 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11555255 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11732524 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29701961 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29701958 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29701956 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29701960 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29701957 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29701959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29702056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29701940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11223487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11360851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701930 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11360852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11223485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11773381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11465198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11773386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11406667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11361094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11465193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29701957 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10416295 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416297 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416301 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416294 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416296 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416293 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416299 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416298 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416292 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416300 - -
          Affymetrix HT MG-430 PM Array Plate 1428478_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10416290 - -
          Mouse Genome 430 2.0 Array 1428478_PM_at - -
          Mouse Genome 430 2.0 Array 1428478_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72549_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72549_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400008034.mm.1 NM_180588.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008034.mm.1 NM_180588.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008034.mm.1 TC1400001000.mm.1 MTA TC
          Mouse Expression Set 430 1428478_PM_at - -
          Mouse Expression Set 430 1428478_at - -
          Affymetrix HT Mouse Genome 430B Array 1428478_at - -
          Affymetrix HT Mouse Genome 430B Array 1428478_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 687426 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4760075 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004760075 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2739250 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2739250 - -
          mouse-6 v1.1 (Illumina) scl46123.6.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 4760075 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46123.6.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4760075 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2739250 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2739250 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline