| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800021454.mm.1 |
NM_180958.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021454.mm.1 |
NM_180958.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021454.mm.1 |
TC0800002803.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800021449.mm.1 |
NM_180958.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021449.mm.1 |
NM_180958.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021449.mm.1 |
TC0800002803.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800021452.mm.1 |
NM_180958.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021452.mm.1 |
NM_180958.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021452.mm.1 |
TC0800002803.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800021466.mm.1 |
NM_180958.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021466.mm.1 |
NM_180958.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021466.mm.1 |
TC0800002803.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800021456.mm.1 |
NM_180958.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021456.mm.1 |
NM_180958.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021456.mm.1 |
TC0800002803.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800021457.mm.1 |
NM_180958.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021457.mm.1 |
NM_180958.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021457.mm.1 |
TC0800002803.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800021460.mm.1 |
NM_180958.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021460.mm.1 |
NM_180958.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021460.mm.1 |
TC0800002803.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800021448.mm.1 |
NM_180958.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021448.mm.1 |
NM_180958.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021448.mm.1 |
TC0800002803.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800021450.mm.1 |
NM_180958.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021450.mm.1 |
NM_180958.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021450.mm.1 |
TC0800002803.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800021461.mm.1 |
NM_180958.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021461.mm.1 |
NM_180958.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021461.mm.1 |
TC0800002803.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800021462.mm.1 |
NM_180958.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021462.mm.1 |
NM_180958.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021462.mm.1 |
TC0800002803.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800021455.mm.1 |
NM_180958.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021455.mm.1 |
NM_180958.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021455.mm.1 |
TC0800002803.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800021458.mm.1 |
NM_180958.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021458.mm.1 |
NM_180958.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021458.mm.1 |
TC0800002803.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800021459.mm.1 |
NM_180958.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021459.mm.1 |
NM_180958.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021459.mm.1 |
TC0800002803.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800021451.mm.1 |
NM_180958.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021451.mm.1 |
NM_180958.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021451.mm.1 |
TC0800002803.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800021463.mm.1 |
NM_180958.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021463.mm.1 |
NM_180958.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800021463.mm.1 |
TC0800002803.mm.1 |
MTA TC |