Search
Search
| Gene Symbol: Fasn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007988.3 | NP_032014.3 | 3 - 4 | 805 | 67 | |
| GenBank mRNA | AF127033.1 | - | 3 - 4 | 322 | 67 | |
| AK147214.1 | - | 3 - 4 | 501 | 67 | ||
| AK147694.1 | - | 3 - 4 | 568 | 67 | ||
| AK171499.1 | - | 3 - 4 | 437 | 67 | ||
| BC046513.1 | - | 3 - 4 | 359 | 4196969 | 67 | |
Set Membership: |
|