Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00663052_g1
          See other PSMD2 GE Assays ›
          Gene Symbol:
          Psmd2
          Gene Name:
          proteasome (prosome, macropain) 26S subunit, non-ATPase, 2
          Gene Aliases:
          9430095H01Rik, AA407121, TEG-190, Tex190
          Chromosome Location:
          Chr.16: 20651652 - 20663414 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00663052_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psmd2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134101.2 NP_598862.1 2 - 3 252 118
          NR_027485.1 2 - 3 344 210
          GenBank mRNA AK146482.1 - 2 - 3 201 118
          AK150374.1 - 2 - 3 224 118
          AK151588.1 - 2 - 3 224 118
          AK152007.1 - 2 - 3 224 118
          AK152970.1 - 2 - 3 221 118
          AK159098.1 - 2 - 3 204 118
          AK159636.1 - 2 - 3 201 118
          AK159756.1 - 2 - 3 224 118
          AK159815.1 - 2 - 3 220 118
          AK166816.1 - 2 - 3 204 118
          AK167326.1 - 2 - 3 204 118
          AK167774.1 - 2 - 3 201 118
          AK167786.1 - 2 - 3 202 118
          AK167909.1 - 2 - 3 212 118
          AK167995.1 - 2 - 3 220 118
          AK168131.1 - 2 - 3 211 118
          AK168398.1 - 2 - 3 204 118
          AK168543.1 - 2 - 3 201 118
          AK169867.1 - 2 - 3 204 118
          AK212329.1 - 2 - 3 80 118
          BC021508.1 - 2 - 3 221 5347139 118
          BC023049.1 - 2 - 3 169 5356268 118
          BC024058.1 - 2 - 3 194 118
          BC024830.1 - 2 - 3 192 5358424 118
          BC027388.1 - 2 - 3 216 4952935 118
          BC027767.1 - 2 - 3 220 5347281 118
          BC027822.1 - 2 - 3 203 5321505 118
          BC028851.1 - 2 - 3 192 3660169 118
          BC031128.1 - 2 - 3 221 4982313 118
          BC033325.1 - 2 - 3 281 210
          BY307723.1 - 2 - 3 253 118

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC023049.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psmd2

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein catabolic process
          protein binding
          enzyme regulator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30588732 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30588728 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30588730 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30588727 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2022987 BC033325.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2022987 BC028851.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2022987 BC031128.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2022987 BC027767.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2022987 BC027822.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252759 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 773354 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 773878 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 19735 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 613306 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 214050 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42378 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123172 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163217 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2022987 BC033325.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2022987 BC028851.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2022987 BC031128.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2022987 BC027767.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2022987 BC027822.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P363920 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P163978 BC033325.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P163978 BC028851.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P163978 BC031128.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P163978 BC027767.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P163978 BC027822.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30588725 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163217 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123172 - -
          Whole Mouse Genome G4122A A_52_P363920 - -
          Whole Mouse Genome G4122A A_52_P163978 BC033325.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P163978 BC028851.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P163978 BC031128.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P163978 BC027767.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P163978 BC027822.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107155 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106684 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112338 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163217 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123172 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12736399 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12672903 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12736388 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12736396 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30588732 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30588727 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30588728 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30588730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30588722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12736390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12521384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12500104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12736383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588701 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13003675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12482730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12764501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12520032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588767 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12523208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588745 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13017438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12736388 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12742365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12603556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30588762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12521383 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10434478 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434468 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434474 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434479 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434475 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434480 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362393 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434471 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434472 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434473 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434470 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434477 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434469 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434476 - -
          Affymetrix HT MG-430 PM Array Plate 1443477_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1415831_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10362392 - -
          Affymetrix Mouse Gene 1.1 ST Array 10434467 - -
          Mouse Genome 430 2.0 Array 1443477_at - -
          Mouse Genome 430 2.0 Array 1415831_at - -
          Mouse Genome 430 2.0 Array 1415831_PM_at - -
          Mouse Genome 430 2.0 Array 1443477_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21762_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21762_at - -
          Affymetrix Mouse Clariom S Array PSR1600002756.mm.1 NM_134101.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600002756.mm.1 NM_134101.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600002756.mm.1 NR_027485.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600002756.mm.1 NR_027485.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600002756.mm.1 TC1600000286.mm.1 MTA TC
          Mouse Expression Set 430 1415831_at - -
          Mouse Expression Set 430 1415831_PM_at - -
          Mouse Expression Set 430 1443477_PM_at - -
          Mouse Expression Set 430 1443477_at - -
          Murine Genome U74v2 Set 92645_PM_at - -
          Murine Genome U74v2 Set 92645_at - -
          Mouse Genome 430A 2.0 Array 1415831_at - -
          Mouse Genome 430A 2.0 Array 1415831_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1415831_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1415831_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 904594 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5050364 - -
          MouseWG-6 v2 Expression BeadChip 4670706 - -
          Illumina mouse-6 v1.0 expression beadchip 4670706 - -
          Illumina mouse-6 v1.0 expression beadchip 5050364 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005050364 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004670706 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2682947 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2682947 - -
          mouse-6 v1.1 (Illumina) scl021762.20_1-S - -
          mouse-6 v1.1 (Illumina) scl0021762.1_119-S - -
          Illumina Sentrix mouse-6.v1 bead array 4670706 - -
          Illumina Sentrix mouse-6.v1 bead array 5050364 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl021762.20_1-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0021762.1_119-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4670706 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5050364 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2682945 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2682947 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2682947 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2741950 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2682945 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0021762.1_119-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl021762.20_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline