Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00663210_mH
          See other C1S1 GE Assays ›
          Gene Symbol:
          C1s1
          Gene Name:
          complement component 1, s subcomponent 1
          Gene Aliases:
          AA959438, AI255193, AI327365, C1s, C1sa
          Chromosome Location:
          Chr.6: 124530344 - 124542390 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00663210_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: C1s1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001097617.1 NP_001091086.1 11 - 12 1512 115
          NM_144938.2 NP_659187.2 11 - 12 1549 115
          GenBank mRNA AF459019.1 - 11 - 12 1476 115
          AK087522.1 - 11 - 12 1481 115
          AK172453.1 - 11 - 12 1477 115
          BC018319.1 - 11 - 12 1441 4196654 115
          BC022123.1 - 11 - 12 1502 5097206 115
          BC111880.1 - 1305 40059749 115

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003108.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene C1s1

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          proteolysis
          complement activation
          complement activation, classical pathway
          innate immune response
          serine-type endopeptidase activity
          calcium ion binding
          peptidase activity
          serine-type peptidase activity
          hydrolase activity
          identical protein binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042612 BC022123.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042612 BC111880.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042612 AK172453.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042612 BC018319.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2042612 AF459019.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 122253 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 703574 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 50953 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042612 BC022123.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042612 BC111880.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042612 AK172453.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042612 BC018319.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2042612 AF459019.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P208793 BC022123.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P208793 BC111880.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P208793 AK172453.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P208793 BC018319.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P208793 AF459019.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P634218 BC022123.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P634218 BC111880.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P634218 AK172453.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P634218 BC018319.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P634218 AF459019.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24815846 - -
          Whole Mouse Genome G4122A A_51_P208793 BC022123.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208793 BC111880.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208793 AK172453.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208793 BC018319.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208793 AF459019.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P634218 BC022123.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P634218 BC111880.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P634218 AK172453.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P634218 BC018319.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P634218 AF459019.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106301 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115448 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05313638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24815818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24815846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24815791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24815858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24815837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24815821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24815792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24815839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24815830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24815818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24815851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24815854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24815841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24815842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24815827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24815784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24815807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05875149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24815845 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10547741 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547746 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547750 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547742 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547743 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547744 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547745 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547748 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547749 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547747 - -
          Affymetrix Mouse Gene 1.1 ST Array 10547740 - -
          Mouse Genome 430 2.0 Array 1424041_PM_s_at - -
          Mouse Genome 430 2.0 Array 1424041_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 50908_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 50908_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600024013.mm.1 NM_001097617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024013.mm.1 NM_001097617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024013.mm.1 NM_144938.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024013.mm.1 NM_144938.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024013.mm.1 TC0600003108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024009.mm.1 NM_001097617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024009.mm.1 NM_001097617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024009.mm.1 NM_144938.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024009.mm.1 NM_144938.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024009.mm.1 TC0600003108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024007.mm.1 NM_001097617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024007.mm.1 NM_001097617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024007.mm.1 NM_144938.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024007.mm.1 NM_144938.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024007.mm.1 TC0600003108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024010.mm.1 NM_001097617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024010.mm.1 NM_001097617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024010.mm.1 NM_144938.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024010.mm.1 NM_144938.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024010.mm.1 TC0600003108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024017.mm.1 NM_001097617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024017.mm.1 NM_001097617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024017.mm.1 NM_144938.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024017.mm.1 NM_144938.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024017.mm.1 TC0600003108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024019.mm.1 NM_001097617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024019.mm.1 NM_001097617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024019.mm.1 NM_144938.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024019.mm.1 NM_144938.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024019.mm.1 TC0600003108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024008.mm.1 NM_001097617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024008.mm.1 NM_001097617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024008.mm.1 NM_144938.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024008.mm.1 NM_144938.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024008.mm.1 TC0600003108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024011.mm.1 NM_001097617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024011.mm.1 NM_001097617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024011.mm.1 NM_144938.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024011.mm.1 NM_144938.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024011.mm.1 TC0600003108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024021.mm.1 NM_001097617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024021.mm.1 NM_001097617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024021.mm.1 NM_144938.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024021.mm.1 NM_144938.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024021.mm.1 TC0600003108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024016.mm.1 NM_001097617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024016.mm.1 NM_001097617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024016.mm.1 NM_144938.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024016.mm.1 NM_144938.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024016.mm.1 TC0600003108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024015.mm.1 NM_001097617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024015.mm.1 NM_001097617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024015.mm.1 NM_144938.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024015.mm.1 NM_144938.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024015.mm.1 TC0600003108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024025.mm.1 NM_144938.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024025.mm.1 NM_144938.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024025.mm.1 TC0600003108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024031.mm.1 NM_001097617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024031.mm.1 NM_001097617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024031.mm.1 TC0600003108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024020.mm.1 NM_001097617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024020.mm.1 NM_001097617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024020.mm.1 NM_144938.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024020.mm.1 NM_144938.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024020.mm.1 TC0600003108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024022.mm.1 NM_001097617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024022.mm.1 NM_001097617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024022.mm.1 NM_144938.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024022.mm.1 NM_144938.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024022.mm.1 TC0600003108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024012.mm.1 NM_001097617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024012.mm.1 NM_001097617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024012.mm.1 NM_144938.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024012.mm.1 NM_144938.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024012.mm.1 TC0600003108.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024018.mm.1 NM_001097617.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024018.mm.1 NM_001097617.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024018.mm.1 NM_144938.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024018.mm.1 NM_144938.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024018.mm.1 TC0600003108.mm.1 MTA TC
          Mouse Expression Set 430 1424041_s_at - -
          Mouse Expression Set 430 1424041_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1424041_s_at - -
          Mouse Genome 430A 2.0 Array 1424041_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 840184 - -
          Illumina mouse-6 v1.0 expression beadchip 840184 - -
          Illumina mouse-6 v1.0 expression beadchip 6840114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000840184 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006840114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1257046 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2691327 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2691327 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257046 - -
          mouse-6 v1.1 (Illumina) scl0050908.1_300-S - -
          mouse-6 v1.1 (Illumina) scl0050908.1_128-S - -
          Illumina Sentrix mouse-6.v1 bead array 840184 - -
          Illumina Sentrix mouse-6.v1 bead array 6840114 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0050908.1_128-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0050908.1_300-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6840114 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 840184 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1257046 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2667269 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257046 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2667269 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline