Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00663500_s1
          See other LDHAL6B GE Assays ›
          Gene Symbol:
          Ldhal6b
          Gene Name:
          lactate dehydrogenase A-like 6B
          Gene Aliases:
          4933402O15Rik, AI326310, LDHL, Ldhal
          Chromosome Location:
          Chr.17: 5417323 - 5418767 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00663500_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ldhal6b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175349.2 NP_780558.1 1 1132 102
          GenBank mRNA AK077089.1 - 1 1078 102
          AY280363.1 - 1 1132 102
          BC061193.1 - 1 1077 6704800 102

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ldhal6b

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P352357 BC061193.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P352357 NM_175349.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P352357 AK077089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P352357 AY280363.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P352357 BC061193.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P352357 NM_175349.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P352357 AK077089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P352357 AY280363.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162919 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P352357 BC061193.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P352357 NM_175349.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P352357 AK077089.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P352357 AY280363.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P352357 BC061193.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P352357 NM_175349.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P352357 AK077089.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P352357 AY280363.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P352357 BC061193.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P352357 NM_175349.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P352357 AK077089.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P352357 AY280363.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P352357 BC061193.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P352357 NM_175349.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P352357 AK077089.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P352357 AY280363.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016888 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016895 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016893 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31016876 - -
          Whole Mouse Genome G4122A A_51_P352357 BC061193.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P352357 NM_175349.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P352357 AK077089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P352357 AY280363.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P352357 BC061193.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P352357 NM_175349.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P352357 AK077089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P352357 AY280363.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13262710 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13262715 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13262682 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13262690 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13554683 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13554697 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13554692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31016911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13262700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016856 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13347073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13413460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13212848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13262689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13212846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13212844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31016906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13548686 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10447558 - -
          Affymetrix Mouse Gene 1.0 ST Array 10447559 - -
          Affymetrix HT MG-430 PM Array Plate 1434247_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10447557 - -
          Mouse Genome 430 2.0 Array 1434247_PM_at - -
          Mouse Genome 430 2.0 Array 1434247_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 106557_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 106557_at - -
          Affymetrix Mouse Clariom S Array PSR1700012358.mm.1 NM_175349.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012358.mm.1 NM_175349.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012358.mm.1 TC1700001367.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012357.mm.1 NM_175349.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012357.mm.1 NM_175349.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012357.mm.1 TC1700001367.mm.1 MTA TC
          Mouse Expression Set 430 1434247_at - -
          Mouse Expression Set 430 1434247_PM_at - -
          Murine Genome U74v2 Set 165429_PM_at - -
          Murine Genome U74v2 Set 165429_at - -
          Affymetrix HT Mouse Genome 430B Array 1434247_at - -
          Affymetrix HT Mouse Genome 430B Array 1434247_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 359903 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5130280 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005130280 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2666402 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2666402 - -
          mouse-6 v1.1 (Illumina) scl0106557.1_261-S - -
          Illumina Sentrix mouse-6.v1 bead array 5130280 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0106557.1_261-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5130280 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0