Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00663741_s1
          See other TAS2R136 GE Assays ›
          Gene Symbol:
          Tas2r136
          Gene Name:
          taste receptor, type 2, member 136
          Gene Aliases:
          Tas2r36, mt2r52
          Chromosome Location:
          Chr.6: 132777179 - 132778162 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00663741_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tas2r136
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181276.1 NP_851793.1 1 943 68

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tas2r136

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          signal transduction
          G-protein coupled receptor signaling pathway
          response to stimulus
          sensory perception of taste
          signal transducer activity
          G-protein coupled receptor activity
          taste receptor activity
          bitter taste receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P102271 NM_181276.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2024120 NM_181276.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 914009 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162010 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P102271 NM_181276.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2024120 NM_181276.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P184480 NM_181276.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24854660 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162010 - -
          Whole Mouse Genome G4122A A_52_P184480 NM_181276.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102271 NM_181276.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162010 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05302453 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05302478 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05302470 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05302469 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05302468 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05302461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24854692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05559616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854667 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05302471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05537680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05112041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05573660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05573663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05302465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05753465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05357363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05001067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05821494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05569089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24854615 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10548668 - -
          Affymetrix Mouse Gene 1.1 ST Array 10548667 - -
          Affymetrix Mouse Clariom S Array PSR0600025833.mm.1 TC0600003288.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600025833.mm.1 NM_181276.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600025833.mm.1 NM_181276.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 382729 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3130458 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003130458 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1250482 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1250482 - -
          mouse-6 v1.1 (Illumina) scl710.1.1_252-S - -
          Illumina Sentrix mouse-6.v1 bead array 3130458 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl710.1.1_252-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3130458 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1250482 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1250482 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline