Search
Search
| Gene Symbol: Fn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276408.1 | NP_001263337.1 | 41 - 42 | 6992 | 71 | |
| NM_001276409.1 | NP_001263338.1 | 41 - 42 | 6917 | 71 | ||
| NM_001276410.1 | NP_001263339.1 | 40 - 41 | 6722 | 71 | ||
| NM_001276411.1 | NP_001263340.1 | 40 - 41 | 6647 | 71 | ||
| NM_001276412.1 | NP_001263341.1 | 41 - 42 | 6632 | 71 | ||
| NM_001276413.1 | NP_001263342.1 | 40 - 41 | 6362 | 71 | ||
| NM_010233.2 | NP_034363.1 | 42 - 43 | 7265 | 71 | ||
| GenBank mRNA | AK090130.1 | - | 42 - 43 | 7217 | 71 | |
| AK090135.1 | - | 42 - 43 | 7205 | 71 | ||
| AK133868.1 | - | 16 - 17 | 2747 | 71 | ||
| AK147249.1 | - | 32 - 33 | 5342 | 71 | ||
| AK147315.1 | - | 41 - 42 | 6856 | 71 | ||
| AK147639.1 | - | 41 - 42 | 6571 | 71 | ||
| AK147663.1 | - | 40 - 41 | 6585 | 71 | ||
| AK147683.1 | - | 41 - 42 | 6931 | 71 | ||
| AK147701.1 | - | 40 - 41 | 6300 | 71 | ||
| AK159794.1 | - | 13 - 14 | 2914 | 71 | ||
| AK170755.1 | - | 40 - 41 | 6690 | 71 | ||
| BC004724.1 | - | 2 - 3 | 230 | 71 | ||
| BC025521.1 | - | 12 - 13 | 2013 | 71 | ||
| BC036167.1 | - | 15 - 16 | 2444 | 71 | ||
| BC085240.1 | - | 13 - 14 | 2332 | 71 | ||
| BC138421.1 | - | 40 - 41 | 6511 | 8861441 | 71 | |
| BC145271.1 | - | 40 - 41 | 6226 | 9053825 | 71 | |
| CN532664.1 | - | 3 - 4 | 346 | 71 | ||
| X93167.1 | - | 25 - 26 | 4320 | 71 | ||
Set Membership: |
|