Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00712273_m1
          See other ARL6IP6 GE Assays ›
          Gene Symbol:
          Arl6ip6
          Gene Name:
          ADP-ribosylation factor-like 6 interacting protein 6
          Gene Aliases:
          2310057C01Rik, 2610529A11Rik, Aip-6
          Chromosome Location:
          Chr.2: 53192084 - 53219221 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00712273_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arl6ip6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022989.4 NP_075365.3 3 - 4 625 70
          GenBank mRNA AK011070.1 - 3 - 4 579 70
          AK016267.1 - 3 - 4 600 70
          AK032740.1 - 3 - 4 599 70
          AK035521.1 - 3 - 4 622 70
          AK052010.1 - 3 - 4 599 70
          AK082824.1 - 3 - 4 597 70
          AK146750.1 - 636 70
          AK159990.1 - 3 - 4 587 70
          BC019550.1 - 3 - 4 592 4210432 70
          BC157980.1 - 3 - 4 616 9088139 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000843.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arl6ip6

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200006905.mm.1 NM_022989.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006905.mm.1 NM_022989.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006905.mm.1 TC0200000843.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200006903.mm.1 NM_022989.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006903.mm.1 NM_022989.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006903.mm.1 TC0200000843.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200006907.mm.1 NM_022989.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006907.mm.1 NM_022989.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006907.mm.1 TC0200000843.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200006904.mm.1 NM_022989.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006904.mm.1 NM_022989.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006904.mm.1 TC0200000843.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200006906.mm.1 NM_022989.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006906.mm.1 NM_022989.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006906.mm.1 TC0200000843.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200006908.mm.1 NM_022989.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006908.mm.1 NM_022989.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006908.mm.1 TC0200000843.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200006902.mm.1 NM_022989.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006902.mm.1 NM_022989.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006902.mm.1 TC0200000843.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200006909.mm.1 NM_022989.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006909.mm.1 NM_022989.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006909.mm.1 TC0200000843.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 925369 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0