Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00712503_m1
          See other MRPL52 GE Assays ›
          Gene Symbol:
          Mrpl52
          Gene Name:
          mitochondrial ribosomal protein L52
          Gene Aliases:
          1110047B07Rik
          Chromosome Location:
          Chr.14: 54426909 - 54429750 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00712503_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrpl52
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026851.2 NP_081127.1 4 - 5 244 67
          GenBank mRNA AK004194.1 - 4 - 5 245 67
          AK150806.1 - 4 - 5 244 67
          AK151615.1 - 4 - 5 244 67
          AK152876.1 - 4 - 5 231 67
          AK164629.1 - 4 - 5 242 67
          BC028522.1 - 4 - 5 277 1448458 67
          BC060043.1 - 3 - 4 254 5683366 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000744.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrpl52

          Panther Classification:

          Molecular Function -

          ribosomal protein translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P315111 BC060043.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P315111 NM_026851.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P315111 AK004194.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P315111 AK150806.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P315111 AK151615.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P315111 BC060043.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P315111 NM_026851.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P315111 AK004194.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P315111 AK150806.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P315111 AK151615.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558314 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171361 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P315111 BC060043.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P315111 NM_026851.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P315111 AK004194.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P315111 AK150806.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P315111 AK151615.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P315111 BC060043.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P315111 NM_026851.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P315111 AK004194.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P315111 AK150806.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P315111 AK151615.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P279852 AK081551.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29612370 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171361 - -
          Whole Mouse Genome G4122A A_52_P279852 AK081551.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P315111 BC060043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P315111 NM_026851.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P315111 AK004194.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P315111 AK150806.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P315111 AK151615.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P315111 BC060043.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P315111 NM_026851.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P315111 AK004194.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P315111 AK150806.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P315111 AK151615.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P217440 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102432 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171361 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11369131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29612383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11446795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11256971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11185193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11481800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11779412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11773483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11446796 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11804663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29612356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11369126 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10415050 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415047 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415048 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415049 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415046 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415051 - -
          Affymetrix HT MG-430 PM Array Plate 1460701_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1455366_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1415761_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1415762_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10415045 - -
          Mouse Genome 430 2.0 Array 1455366_PM_at - -
          Mouse Genome 430 2.0 Array 1455366_at - -
          Mouse Genome 430 2.0 Array 1460701_PM_a_at - -
          Mouse Genome 430 2.0 Array 1415762_x_at - -
          Mouse Genome 430 2.0 Array 1415762_PM_x_at - -
          Mouse Genome 430 2.0 Array 1415761_at - -
          Mouse Genome 430 2.0 Array 1460701_a_at - -
          Mouse Genome 430 2.0 Array 1415761_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68836_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68836_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400005807.mm.1 NM_026851.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400005807.mm.1 NM_026851.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400005807.mm.1 TC1400000744.mm.1 MTA TC
          Mouse Expression Set 430 1460701_PM_a_at - -
          Mouse Expression Set 430 1415762_x_at - -
          Mouse Expression Set 430 1415761_at - -
          Mouse Expression Set 430 1455366_at - -
          Mouse Expression Set 430 1415762_PM_x_at - -
          Mouse Expression Set 430 1455366_PM_at - -
          Mouse Expression Set 430 1415761_PM_at - -
          Mouse Expression Set 430 1460701_a_at - -
          Mouse Expression Set 430 1415761_PM_at - -
          Mouse Expression Set 430 1415762_x_at - -
          Mouse Expression Set 430 1415761_at - -
          Mouse Expression Set 430 1415762_PM_x_at - -
          Murine Genome U74v2 Set 97443_at - -
          Murine Genome U74v2 Set 97443_PM_at - -
          Mouse Genome 430A 2.0 Array 1415762_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1415761_at - -
          Mouse Genome 430A 2.0 Array 1460701_a_at - -
          Mouse Genome 430A 2.0 Array 1415762_x_at - -
          Mouse Genome 430A 2.0 Array 1455366_PM_at - -
          Mouse Genome 430A 2.0 Array 1455366_at - -
          Mouse Genome 430A 2.0 Array 1415761_PM_at - -
          Mouse Genome 430A 2.0 Array 1460701_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1415762_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1415761_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1455366_at - -
          Affymetrix HT Mouse Genome 430A Array 1460701_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1455366_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460701_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1415762_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1415761_at - -
          Affymetrix HT Mouse Genome 430B Array 1415761_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1415762_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1415761_at - -
          Affymetrix HT Mouse Genome 430B Array 1415762_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 496861 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1170204 - -
          Illumina mouse-6 v1.0 expression beadchip 1170204 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001170204 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2601865 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2601865 - -
          mouse-6 v1.1 (Illumina) scl46304.5.1_86-S - -
          Illumina Sentrix mouse-6.v1 bead array 1170204 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46304.5.1_86-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170204 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2601865 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2601865 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl46304.5.1_86-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline