Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00717664_m1
          See other GGN GE Assays ›
          Gene Symbol:
          Ggn
          Gene Name:
          gametogenetin
          Gene Aliases:
          AI593290
          Chromosome Location:
          Chr.7: 29170210 - 29173939 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00717664_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ggn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_182694.2 NP_874353.2 2 - 3 2475 97
          NM_182696.2 NP_874355.1 2 - 3 431 97
          GenBank mRNA AF538032.1 - 2 - 3 1964 97
          AF538033.1 - 3 - 4 1384 97
          AF538034.1 - 2 - 3 431 97
          BC089358.1 - 2 - 3 2475 30610755 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000524.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ggn

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair
          protein complex assembly
          multicellular organism development
          spermatogenesis
          embryo implantation
          protein localization
          cell differentiation
          protein binding
          ubiquitin protein ligase binding
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25025750 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25025741 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25025751 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25025752 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25025740 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25025742 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25025744 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25025743 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107239 BC089358.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107239 NM_182694.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2107239 NM_182696.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454751 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 385580 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151505 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107239 BC089358.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107239 NM_182694.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2107239 NM_182696.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25025744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25025745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25025743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25025746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25025739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25025748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25025740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25025754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25025742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25025757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25025747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25025753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25025760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25025749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25025755 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151505 - -
          Whole Mouse Genome G4122A A_52_P401679 BC089358.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P401679 AF538032.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P401679 AF538033.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P401679 AF538034.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P401679 NM_182694.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113176 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151505 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06258476 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05989447 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06258472 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25025740 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25025750 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25025744 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25025751 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25025752 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25025741 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25025742 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25025743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25025708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05989455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05989450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06258477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06258474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06227041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05989456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25025737 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551723 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551717 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551720 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551722 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551721 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551718 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551719 - -
          Affymetrix HT MG-430 PM Array Plate 1445278_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10551716 - -
          Mouse Genome 430 2.0 Array 1445278_at - -
          Mouse Genome 430 2.0 Array 1445278_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 243897_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 243897_at - -
          Affymetrix Mouse Clariom S Array PSR0700004462.mm.1 TC0700000524.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004462.mm.1 NM_182694.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004462.mm.1 NM_182694.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004462.mm.1 NM_182696.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004462.mm.1 NM_182696.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004463.mm.1 TC0700000524.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004463.mm.1 NM_182694.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004463.mm.1 NM_182694.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004463.mm.1 NM_182696.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004463.mm.1 NM_182696.2 Refseq
          Mouse Expression Set 430 1445278_at - -
          Mouse Expression Set 430 1445278_PM_at - -
          Murine Genome U74v2 Set 140653_PM_at - -
          Murine Genome U74v2 Set 140653_at - -
          Affymetrix HT Mouse Genome 430B Array 1445278_at - -
          Affymetrix HT Mouse Genome 430B Array 1445278_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 357179 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1410528 - -
          MouseWG-6 v2 Expression BeadChip 4210082 - -
          Illumina mouse-6 v1.0 expression beadchip 4210082 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004210082 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237951 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2712196 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2712196 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237951 - -
          mouse-6 v1.1 (Illumina) scl000123.1_453-S - -
          Illumina Sentrix mouse-6.v1 bead array 4210082 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000123.1_453-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4210082 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2712196 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1237951 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2712196 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline