Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00723178_m1
          See other ZFP454 GE Assays ›
          Gene Symbol:
          Zfp454
          Gene Name:
          zinc finger protein 454
          Gene Aliases:
          B230342N19
          Chromosome Location:
          Chr.11: 50872715 - 50887635 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00723178_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp454
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172794.2 NP_766382.2 4 - 5 429 80
          GenBank mRNA BC049793.1 - 4 - 5 429 6510760 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002682.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp454

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          transcription factor activity, sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048775 NM_172794.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048775 BC049793.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048769 NM_172794.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048769 AK046117.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2048769 BC049793.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579570 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242770 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884208 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 589198 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 325438 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 45664 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723187 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430692 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 77918 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139184 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124890 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175406 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114390 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048775 NM_172794.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048775 BC049793.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048769 NM_172794.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048769 AK046117.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2048769 BC049793.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27791124 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124890 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175406 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114390 - -
          Whole Mouse Genome G4122A A_51_P351919 NM_172794.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P351919 AK046117.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P351919 BC049793.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117494 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175406 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124890 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114390 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09528964 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09528956 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09528973 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09528972 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09528957 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09528967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27790995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27791093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09545748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09550392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09390093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09528957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09545756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27790995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09528964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09545751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09431342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09151154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09390097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09528974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27791110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09133353 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10385648 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385651 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385654 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385652 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385649 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385653 - -
          Affymetrix Mouse Gene 1.0 ST Array 10385650 - -
          Affymetrix HT MG-430 PM Array Plate 1456507_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10385647 - -
          Mouse Genome 430 2.0 Array 1456507_at - -
          Mouse Genome 430 2.0 Array 1456507_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 237758_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 237758_at - -
          Affymetrix Mouse Clariom S Array PSR1100024469.mm.1 NM_172794.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024469.mm.1 NM_172794.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024469.mm.1 TC1100002682.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100024466.mm.1 NM_172794.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024466.mm.1 NM_172794.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024466.mm.1 TC1100002682.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100024467.mm.1 NM_172794.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024467.mm.1 NM_172794.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024467.mm.1 TC1100002682.mm.1 MTA TC
          Mouse Expression Set 430 1456507_PM_at - -
          Mouse Expression Set 430 1456507_at - -
          Affymetrix HT Mouse Genome 430B Array 1456507_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456507_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 737278 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6840687 - -
          Illumina mouse-6 v1.0 expression beadchip 6840687 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006840687 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2446113 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2446113 - -
          mouse-6 v1.1 (Illumina) scl40267.6_442-S - -
          Illumina Sentrix mouse-6.v1 bead array 6840687 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40267.6_442-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6840687 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1247155 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247155 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0