Search
Search
| Gene Symbol: Sspo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173428.3 | NP_775604.3 | 80 - 81 | 12144 | 67 | |
| GenBank mRNA | AJ491857.1 | - | 77 - 78 | 11715 | 67 | |
| AK142699.1 | - | 2 - 3 | 237 | 67 | ||
Set Membership: |
|