| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P353905 |
NM_173862.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P353905 |
NM_173862.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2085855 |
BC030396.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
733933 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
74219 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
456030 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
824803 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
664492 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
902368 |
AK146116.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
902368 |
X80699.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
902368 |
XM_001478773.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
902368 |
XM_001478782.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
902368 |
XM_001479777.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
902368 |
XM_001479784.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
776566 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
704636 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
208796 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
428259 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
234424 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
895801 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
636617 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
498758 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
364878 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
916186 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
306650 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
741488 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
90516 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
145818 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
100988 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P100436 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P353905 |
NM_173862.2 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P353905 |
NM_173862.2 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2085855 |
BC030396.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245399 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245398 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245414 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245389 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245410 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245387 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245412 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245372 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245364 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245373 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245376 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245391 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245380 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245402 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245408 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245374 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245368 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245406 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245369 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245405 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245383 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245378 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245394 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245362 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245400 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245403 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245385 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245363 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245393 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245366 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245396 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245360 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30245397 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P100436 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03043233 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P243522 |
BC030396.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P243522 |
NM_173862.2 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P136633 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P120747 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P100436 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11937904 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11937906 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11937895 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11937889 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11937901 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245350 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245502 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245541 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245455 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245405 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245375 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245504 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245393 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245354 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245499 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245347 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245449 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245558 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245590 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245498 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245459 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245457 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245351 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245483 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245420 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245581 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245364 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245566 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245481 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245431 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245583 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245342 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245383 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245389 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245586 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245453 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245582 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245514 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245535 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245463 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245542 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245442 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245599 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245387 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245555 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245385 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245422 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245451 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245528 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245564 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245567 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245391 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245503 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245506 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245376 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245522 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245496 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245570 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245509 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245594 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245488 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245416 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245402 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245500 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245548 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245366 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245588 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245553 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245433 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245561 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245373 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245497 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245396 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245427 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245434 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245372 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245480 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245524 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245337 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245399 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245335 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245379 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245460 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245526 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245440 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245356 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245525 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245352 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245394 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245476 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245400 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245584 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245467 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245596 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245484 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245338 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245430 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245493 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245491 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245515 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245334 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245406 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245368 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245519 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245369 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245378 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245426 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245360 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245543 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245556 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245470 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245466 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245486 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245412 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245380 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245578 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245397 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245579 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245577 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245575 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245531 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245444 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245362 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245421 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245552 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245448 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245550 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245408 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30245508 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245546 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245378 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245424 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245568 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245555 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12306650 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245561 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245405 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245366 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245548 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245352 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12152655 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12103142 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12193091 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245566 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245444 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245438 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245542 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245372 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245537 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12193093 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245354 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12360319 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245434 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245347 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245393 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245394 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245421 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245531 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245415 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245526 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245358 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245430 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245418 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245426 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245564 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245556 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12077879 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245408 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12193088 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245357 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245338 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12445992 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245362 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12102033 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245567 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245374 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245425 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245420 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12007340 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245521 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12103141 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245360 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245544 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245402 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245389 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245570 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245412 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245433 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245399 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245427 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12145635 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245410 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245387 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245528 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11937877 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245414 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12007341 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245383 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245385 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245350 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245364 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245431 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245341 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245437 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245422 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12074647 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245391 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245379 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245442 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245356 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245541 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11937882 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245396 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245441 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245543 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245376 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245446 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245339 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245334 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245369 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245375 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30245380 |
- |
- |