Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00723484_m1
          See other ATP2C1 GE Assays ›
          Gene Symbol:
          Atp2c1
          Gene Name:
          ATPase, Ca++-sequestering
          Gene Aliases:
          1700121J11Rik, ATP2C1A, AW061228, BCPM, D930003G21Rik, HHD, SPCA, pmr1
          Chromosome Location:
          Chr.9: 105411362 - 105521257 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00723484_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp2c1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001253831.1 NP_001240760.1 5 - 6 574 117
          NM_001253834.1 NP_001240763.1 6 - 7 665 117
          NM_001253836.1 NP_001240765.1 5 - 6 597 117
          NM_175025.4 NP_778190.3 6 - 7 780 117
          GenBank mRNA AJ551270.1 - 4 - 5 362 117
          AK007226.1 - 7 - 8 566 117
          AK028393.1 - 5 - 6 543 117
          AK133703.1 - 5 - 6 575 117
          AK136062.1 - 665 117
          AK159918.1 - 6 - 7 731 117
          AK163049.1 - 6 - 7 626 117
          BC043091.1 - 6 - 7 746 6405251 117
          BC043706.1 - 6 - 7 727 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002991.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Transporter Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp2c1

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          calcium ion transport
          manganese ion transport
          cellular calcium ion homeostasis
          signal transduction
          epidermis development
          calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
          cellular manganese ion homeostasis
          actin cytoskeleton reorganization
          Golgi calcium ion homeostasis
          Golgi calcium ion transport
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          calcium ion transmembrane transport
          manganese ion transmembrane transport
          nucleotide binding
          signal transducer activity
          calcium-transporting ATPase activity
          calcium ion binding
          ATP binding
          manganese-transporting ATPase activity
          hydrolase activity
          manganese ion binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900024710.mm.1 NM_001253831.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024710.mm.1 NM_001253831.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024710.mm.1 NM_001253834.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024710.mm.1 NM_001253834.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024710.mm.1 NM_001253836.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024710.mm.1 NM_001253836.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024710.mm.1 NM_175025.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024710.mm.1 NM_175025.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024710.mm.1 TC0900002991.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900024711.mm.1 NM_001253831.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024711.mm.1 NM_001253831.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024711.mm.1 NM_001253834.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024711.mm.1 NM_001253834.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024711.mm.1 NM_001253836.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024711.mm.1 NM_001253836.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024711.mm.1 NM_175025.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024711.mm.1 NM_175025.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900024711.mm.1 TC0900002991.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 388850 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline