| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900006242.mm.1 |
NM_001077631.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006242.mm.1 |
NM_175153.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006242.mm.1 |
TC0900000825.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006240.mm.1 |
NM_001077631.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006240.mm.1 |
NM_175153.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006240.mm.1 |
TC0900000825.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006238.mm.1 |
NM_001077631.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006238.mm.1 |
NM_175153.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006238.mm.1 |
TC0900000825.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006239.mm.1 |
NM_001077631.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006239.mm.1 |
NM_175153.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006239.mm.1 |
TC0900000825.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006241.mm.1 |
NM_001077631.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006241.mm.1 |
NM_175153.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006241.mm.1 |
TC0900000825.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900006236.mm.1 |
NM_001077631.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006236.mm.1 |
NM_175153.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900006236.mm.1 |
TC0900000825.mm.1 |
MTA TC |