Search
Search

Ltbp4| Gene Symbol: Ltbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113549.1 | NP_001107021.1 | 22 - 23 | 3394 | 78 | |
| NM_175641.2 | NP_783572.2 | 25 - 26 | 3593 | 78 | ||
| GenBank mRNA | AF410798.1 | - | 25 - 26 | 3674 | 78 | |
| AF410799.1 | - | 22 - 23 | 3370 | 78 | ||
| AK054297.1 | - | 22 - 23 | 3360 | 78 | ||
| AK157031.1 | - | 22 - 23 | 3410 | 78 | ||
| BC032073.1 | - | 23 - 24 | 3346 | 78 | ||
| BC059016.1 | - | 22 - 23 | 3371 | 78 | ||
| BC141005.1 | - | 25 - 26 | 3599 | 9055617 | 78 | |
Set Membership: |
|