Search
Search
| Gene Symbol: Kif6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177052.3 | NP_796026.2 | 4 - 5 | 466 | 100 | |
| GenBank mRNA | AB001434.1 | - | 1 - 2 | 125 | 100 | |
| AK043404.1 | - | 4 - 5 | 423 | 100 | ||
| AK083760.1 | - | 4 - 5 | 503 | 100 | ||
Set Membership: |
|