Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00724014_m1
          See other LRRN4 GE Assays ›
          Gene Symbol:
          Lrrn4
          Gene Name:
          leucine rich repeat neuronal 4
          Gene Aliases:
          B430119L13Rik, Nlrr4
          Chromosome Location:
          Chr.2: 132866194 - 132894009 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00724014_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrn4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177303.4 NP_796277.2 2 - 3 755 56
          GenBank mRNA AK080889.1 - 2 - 3 756 56
          AK085246.1 - 2 - 3 785 56
          AY325108.1 - 2 - 3 738 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004683.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lrrn4

          Panther Classification:

          Molecular Function -

          immunoglobulin superfamily cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          long-term memory
          visual learning
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21642726 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21642728 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21642730 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P460836 NM_177303.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P460836 AK080889.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P460836 NM_177303.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P460836 AK080889.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 327055 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 908173 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900707 AB031055.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900707 AK088903.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900707 AK090079.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900707 BC094631.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 900707 NM_013898.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 935642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 655477 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887719 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 64663 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 27433 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122374 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156149 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P460836 NM_177303.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P460836 AK080889.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P460836 NM_177303.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P460836 AK080889.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21642743 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156149 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122374 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00497008 - -
          Whole Mouse Genome G4122A A_52_P460836 NM_177303.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P460836 AK080889.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P460836 NM_177303.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P460836 AK080889.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113247 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156149 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122374 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02025032 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02025034 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02025012 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02025029 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02025036 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02025026 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21642728 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21642726 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21642730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21642671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02220305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02273934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02025024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01155331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01758072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01574398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02025034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02025026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02049471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01574399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01155325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02049470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01155323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02025029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01929999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01237599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642716 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01930000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01574386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02118394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02118393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21642712 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10487992 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487993 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487988 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487990 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487989 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487991 - -
          Affymetrix HT MG-430 PM Array Plate 1435648_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10487987 - -
          Mouse Genome 430 2.0 Array 1435648_at - -
          Mouse Genome 430 2.0 Array 1435648_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 320974_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 320974_at - -
          Affymetrix Mouse Clariom S Array PSR0200040243.mm.1 NM_177303.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040243.mm.1 NM_177303.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040243.mm.1 TC0200004683.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200040240.mm.1 NM_177303.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040240.mm.1 NM_177303.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040240.mm.1 TC0200004683.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200040242.mm.1 NM_177303.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040242.mm.1 NM_177303.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040242.mm.1 TC0200004683.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200040241.mm.1 NM_177303.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040241.mm.1 NM_177303.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040241.mm.1 TC0200004683.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200040239.mm.1 NM_177303.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040239.mm.1 NM_177303.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040239.mm.1 TC0200004683.mm.1 MTA TC
          Mouse Expression Set 430 1435648_PM_at - -
          Mouse Expression Set 430 1435648_at - -
          Murine Genome U74v2 Set 168031_PM_r_at - -
          Murine Genome U74v2 Set 168031_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1435648_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435648_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 632154 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5890338 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005890338 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2608166 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2608166 - -
          mouse-6 v1.1 (Illumina) scl0320974.1_66-S - -
          Illumina Sentrix mouse-6.v1 bead array 5890338 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0320974.1_66-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5890338 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2608166 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2608166 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline