Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00724087_m1
          See other CRB3 GE Assays ›
          Gene Symbol:
          Crb3
          Gene Name:
          crumbs family member 3
          Gene Aliases:
          5730439B18, AU015179, AU041752
          Chromosome Location:
          Chr.17: 57062096 - 57065920 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00724087_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crb3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177638.4 NP_808306.1 2 - 3 268 77
          GenBank mRNA AK077528.1 - 2 - 3 200 77
          AK167279.1 - 2 - 3 198 77
          BC024462.1 - 2 - 3 268 4975562 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001023.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Crb3

          Panther Classification:

          Molecular Function -

          cell adhesion molecule

          Gene Ontology Categories:

          Function(s)

          establishment of epithelial cell apical/basal polarity
          cell-cell junction organization

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259846 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259850 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259845 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259849 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259848 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137979 AK077528.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137979 AK167279.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137979 NM_177638.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P328089 AK077528.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P328089 AK167279.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P328089 NM_177638.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P328089 BC024462.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P328089 AK077528.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P328089 AK167279.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P328089 NM_177638.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P328089 BC024462.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 46046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 871748 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 619594 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121348 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157246 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171826 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2137979 AK077528.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2137979 AK167279.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2137979 NM_177638.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P328089 AK077528.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P328089 AK167279.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P328089 NM_177638.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P328089 BC024462.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P328089 AK077528.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P328089 AK167279.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P328089 NM_177638.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P328089 BC024462.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P328089 AK077528.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P328089 AK167279.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P328089 NM_177638.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P328089 BC024462.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P328089 AK077528.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P328089 AK167279.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P328089 NM_177638.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P328089 BC024462.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259860 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171826 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157246 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121348 - -
          Whole Mouse Genome G4122A A_51_P328089 AK077528.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328089 AK167279.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328089 NM_177638.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328089 BC024462.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328089 AK077528.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328089 AK167279.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328089 NM_177638.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328089 BC024462.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130345 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171826 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121348 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157246 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13492415 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259846 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259849 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259850 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259848 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13192156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13492412 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13081242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13492414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259850 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10446227 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446226 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446228 - -
          Affymetrix Mouse Gene 1.0 ST Array 10446225 - -
          Affymetrix HT MG-430 PM Array Plate 1434851_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1455115_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1424809_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10446224 - -
          Mouse Genome 430 2.0 Array 1455115_a_at - -
          Mouse Genome 430 2.0 Array 1455115_PM_a_at - -
          Mouse Genome 430 2.0 Array 1434851_PM_s_at - -
          Mouse Genome 430 2.0 Array 1434851_s_at - -
          Mouse Genome 430 2.0 Array 1424809_PM_at - -
          Mouse Genome 430 2.0 Array 1424809_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 224912_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 224912_at - -
          Affymetrix Mouse Clariom S Array PSR1700009871.mm.1 NM_177638.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009871.mm.1 NM_177638.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700009871.mm.1 TC1700001023.mm.1 MTA TC
          Mouse Expression Set 430 1424809_PM_at - -
          Mouse Expression Set 430 1455115_a_at - -
          Mouse Expression Set 430 1455115_PM_a_at - -
          Mouse Expression Set 430 1434851_PM_s_at - -
          Mouse Expression Set 430 1424809_at - -
          Mouse Expression Set 430 1434851_s_at - -
          Murine Genome U74v2 Set 104585_at - -
          Murine Genome U74v2 Set 104584_f_at - -
          Murine Genome U74v2 Set 104585_PM_at - -
          Murine Genome U74v2 Set 104584_PM_f_at - -
          Murine Genome U74v2 Set 168426_f_at - -
          Murine Genome U74v2 Set 168426_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1455115_a_at - -
          Mouse Genome 430A 2.0 Array 1434851_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1434851_s_at - -
          Mouse Genome 430A 2.0 Array 1424809_at - -
          Mouse Genome 430A 2.0 Array 1424809_PM_at - -
          Mouse Genome 430A 2.0 Array 1455115_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1455115_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1424809_at - -
          Affymetrix HT Mouse Genome 430A Array 1434851_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1434851_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1455115_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1424809_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 830412 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6940091 - -
          Illumina mouse-6 v1.0 expression beadchip 6940091 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006940091 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2755888 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2755888 - -
          mouse-6 v1.1 (Illumina) scl50580.5_35-S - -
          Illumina Sentrix mouse-6.v1 bead array 6940091 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50580.5_35-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6940091 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2755888 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2755888 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline