Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00724835_m1
          See other WDR92 GE Assays ›
          Gene Symbol:
          Wdr92
          Gene Name:
          WD repeat domain 92
          Gene Aliases:
          AI553587, HZGJ
          Chromosome Location:
          Chr.11: 17211893 - 17235200 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00724835_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wdr92
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178909.4 NP_849240.1 3 - 4 484 63
          GenBank mRNA AK033294.1 - 3 - 4 430 63
          AK047691.1 - 3 - 4 459 63
          AK088218.1 - 3 - 4 459 63
          AK142228.1 - 3 - 4 430 63
          BC096540.1 - 3 - 4 413 6403681 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100004262.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Wdr92

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          histone lysine methylation
          methylated histone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2172309 NM_178909.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071952 AK139758.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751728 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 181472 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 557177 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 353645 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 245348 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 793050 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 720127 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20397 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 748755 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 39057 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 677666 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 76086 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 19908 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170477 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143142 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105109 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071952 AK139758.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2172309 NM_178909.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27614825 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143142 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07265367 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105109 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07265393 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07265361 - -
          Whole Mouse Genome G4122A A_51_P208862 AK088218.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208862 AK142228.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208862 BC096540.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208862 NM_178909.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P657759 AK088218.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P657759 AK142228.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P657759 BC096540.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P657759 NM_178909.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P657759 AK033294.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143142 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105109 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09445329 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09445334 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09445331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27615012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27615007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27615005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27615018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27615015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27615001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27614826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27615002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09718594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09000642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09024104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09130913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09130902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09718581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09446336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09619249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09129067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09408356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09256944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09129065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27614837 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10374434 - -
          Affymetrix Mouse Gene 1.0 ST Array 10374435 - -
          Affymetrix Mouse Gene 1.0 ST Array 10374432 - -
          Affymetrix Mouse Gene 1.0 ST Array 10374433 - -
          Affymetrix Mouse Gene 1.0 ST Array 10374438 - -
          Affymetrix Mouse Gene 1.0 ST Array 10374437 - -
          Affymetrix Mouse Gene 1.0 ST Array 10374436 - -
          Affymetrix Mouse Gene 1.0 ST Array 10374441 - -
          Affymetrix Mouse Gene 1.0 ST Array 10374439 - -
          Affymetrix Mouse Gene 1.0 ST Array 10374440 - -
          Affymetrix HT MG-430 PM Array Plate 1454922_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1458355_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1447230_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10374430 - -
          Mouse Genome 430 2.0 Array 1454922_PM_at - -
          Mouse Genome 430 2.0 Array 1458355_x_at - -
          Mouse Genome 430 2.0 Array 1447230_at - -
          Mouse Genome 430 2.0 Array 1454922_at - -
          Mouse Genome 430 2.0 Array 1458355_PM_x_at - -
          Mouse Genome 430 2.0 Array 1447230_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 103784_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 103784_at - -
          Affymetrix Mouse Clariom S Array PSR1100001525.mm.1 NM_178909.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100001525.mm.1 NM_178909.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100001525.mm.1 TC1100004262.mm.1 MTA TC
          Mouse Expression Set 430 1454922_PM_at - -
          Mouse Expression Set 430 1447230_at - -
          Mouse Expression Set 430 1458355_x_at - -
          Mouse Expression Set 430 1447230_PM_at - -
          Mouse Expression Set 430 1454922_at - -
          Mouse Expression Set 430 1458355_PM_x_at - -
          Murine Genome U74v2 Set 99849_PM_at - -
          Murine Genome U74v2 Set 99849_at - -
          Affymetrix HT Mouse Genome 430B Array 1454922_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447230_at - -
          Affymetrix HT Mouse Genome 430B Array 1458355_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1454922_at - -
          Affymetrix HT Mouse Genome 430B Array 1458355_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1447230_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 583854 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5570097 - -
          Illumina mouse-6 v1.0 expression beadchip 5570097 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005570097 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1217295 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225370 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1217295 - -
          mouse-6 v1.1 (Illumina) scl0103784.8_117-S - -
          Illumina Sentrix mouse-6.v1 bead array 5570097 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0103784.8_117-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5570097 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2615448 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2615448 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline