Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00724869_m1
          See other PDRG1 GE Assays ›
          Gene Symbol:
          Pdrg1
          Gene Name:
          p53 and DNA damage regulated 1
          Gene Aliases:
          1110004D19Rik, AU020945, AV006589, Pdrg
          Chromosome Location:
          Chr.2: 153008890 - 153015383 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00724869_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pdrg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178939.2 NP_849270.1 1 - 2 150 90
          GenBank mRNA AK003410.1 - 1 - 2 152 90
          AK145939.1 - 1 - 2 195 90
          AY286302.1 - 1 - 2 131 90
          BC031699.1 - 1 - 2 150 4020218 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004922.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pdrg1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          biological_process
          molecular_function
          unfolded protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21752415 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21752417 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21752418 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21752416 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P173961 AK145939.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P173961 AY286302.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P173961 NM_178939.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P173961 AK003410.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P173961 AK145939.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P173961 AY286302.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P173961 NM_178939.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P173961 AK003410.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207311 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 835980 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 101734 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600869 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 105010 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124613 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P173961 AK145939.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P173961 AY286302.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P173961 NM_178939.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P173961 AK003410.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P173961 AK145939.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P173961 AY286302.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P173961 NM_178939.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P173961 AK003410.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752425 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21752421 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124613 - -
          Whole Mouse Genome G4122A A_51_P173961 AK145939.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P173961 AY286302.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P173961 NM_178939.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P173961 AK003410.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P173961 AK145939.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P173961 AY286302.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P173961 NM_178939.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P173961 AK003410.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130080 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124613 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02154694 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01677123 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01388023 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01677125 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02154696 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21752417 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21752418 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21752416 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21752415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21752393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02181495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752415 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752401 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01712917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02269258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02234892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02181492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01712919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02269260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02181488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01715557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02133689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02181503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02154694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02181489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01715558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02234893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02181501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02269263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02234899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02181509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02234660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02181490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02154696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02133690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02234898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01388023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02234887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02181494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01712916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01810546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01451273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752460 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01953803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01451272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21752430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02234901 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10488688 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488690 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488689 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488692 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488691 - -
          Affymetrix HT MG-430 PM Array Plate 1419933_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1428308_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1428309_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10488687 - -
          Mouse Genome 430 2.0 Array 1428308_PM_at - -
          Mouse Genome 430 2.0 Array 1419934_at - -
          Mouse Genome 430 2.0 Array 1419933_at - -
          Mouse Genome 430 2.0 Array 1428308_at - -
          Mouse Genome 430 2.0 Array 1428309_PM_s_at - -
          Mouse Genome 430 2.0 Array 1428309_s_at - -
          Mouse Genome 430 2.0 Array 1419933_PM_at - -
          Mouse Genome 430 2.0 Array 1419934_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68559_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68559_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200041740.mm.1 NM_178939.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041740.mm.1 NM_178939.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041740.mm.1 TC0200004922.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041736.mm.1 NM_178939.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041736.mm.1 NM_178939.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041736.mm.1 TC0200004922.mm.1 MTA TC
          Mouse Expression Set 430 1419933_PM_at - -
          Mouse Expression Set 430 1428309_PM_s_at - -
          Mouse Expression Set 430 1419933_at - -
          Mouse Expression Set 430 1428308_at - -
          Mouse Expression Set 430 1428309_s_at - -
          Mouse Expression Set 430 1428308_PM_at - -
          Murine Genome U74v2 Set 92806_at - -
          Murine Genome U74v2 Set 92806_PM_at - -
          Mouse Genome 430A 2.0 Array 1419933_at - -
          Mouse Genome 430A 2.0 Array 1419933_PM_at - -
          Mouse Genome 430A 2.0 Array 1428308_at - -
          Mouse Genome 430A 2.0 Array 1428309_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1428309_s_at - -
          Mouse Genome 430A 2.0 Array 1428308_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428308_at - -
          Affymetrix HT Mouse Genome 430A Array 1428309_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1428309_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1428308_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419933_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419933_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 829551 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5340592 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005340592 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1249417 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249417 - -
          mouse-6 v1.1 (Illumina) scl068559.1_22-S - -
          Illumina Sentrix mouse-6.v1 bead array 5340592 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl068559.1_22-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5340592 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1249417 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1249417 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl068559.1_22-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline