Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00725148_m1
          See other DEFB37 GE Assays ›
          Gene Symbol:
          Defb37
          Gene Name:
          defensin beta 37
          Gene Aliases:
          BD-37
          Chromosome Location:
          Chr.8: 18986233 - 18991055 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00725148_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Defb37
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181683.2 NP_859011.1 1 - 2 144 86
          GenBank mRNA AJ489588.1 - 1 - 2 144 86
          AJ578470.1 - 1 - 2 62 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001798.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Defb37

          Panther Classification:

          Molecular Function -

          antimicrobial response protein defense/immunity protein

          Gene Ontology Categories:

          Function(s) Process(es)

          defense response
          biological_process
          defense response to bacterium
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P229791 AJ489588.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P229791 AJ489588.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 267462 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 213495 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125179 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P229791 AJ489588.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P229791 AJ489588.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25701701 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125179 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06442990 - -
          Whole Mouse Genome G4122A A_52_P229791 AJ489588.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P229791 AJ489588.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132933 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125179 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06735189 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06735196 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06735198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25701701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06939109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701734 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07125218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06924108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06888569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07360561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25701692 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10577341 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577342 - -
          Affymetrix Mouse Gene 1.1 ST Array 10577340 - -
          Affymetrix Mouse Clariom S Array PSR0800014351.mm.1 NM_181683.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014351.mm.1 NM_181683.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014351.mm.1 TC0800001798.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014353.mm.1 NM_181683.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014353.mm.1 NM_181683.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014353.mm.1 TC0800001798.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014354.mm.1 NM_181683.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014354.mm.1 NM_181683.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014354.mm.1 TC0800001798.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014352.mm.1 NM_181683.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014352.mm.1 NM_181683.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014352.mm.1 TC0800001798.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 400148 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 520592 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000520592 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237610 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237610 - -
          mouse-6 v1.1 (Illumina) scl35038.2.1_104-S - -
          Illumina Sentrix mouse-6.v1 bead array 520592 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35038.2.1_104-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 520592 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2913432 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2913432 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2770363 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0