Search
Search
| Gene Symbol: Twf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008971.4 | NP_032997.3 | 9 | 1032 | 82 | |
| GenBank mRNA | AK077410.1 | - | 9 | 1091 | 82 | |
| AK147990.1 | - | 9 | 1026 | 82 | ||
| AK150852.1 | - | 9 | 1055 | 82 | ||
| AK168142.1 | - | 9 | 1050 | 82 | ||
| AK168161.1 | - | 9 | 1050 | 82 | ||
| AY267188.1 | - | 9 | 984 | 82 | ||
| BC015081.1 | - | 9 | 1049 | 3979221 | 82 | |
| BC094034.1 | - | 9 | 1032 | 4934593 | 82 | |
| U82324.1 | - | 9 | 1040 | 82 | ||
Set Membership: |
|