Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00726185_s1
          See other TUBB4A GE Assays ›
          Gene Symbol:
          Tubb4a
          Gene Name:
          tubulin, beta 4A class IVA
          Gene Aliases:
          AI325297, M(beta)4, Tubb, Tubb4
          Chromosome Location:
          Chr.17: 57080066 - 57087782 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00726185_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tubb4a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009451.3 NP_033477.2 4 1941 89
          GenBank mRNA AK013717.1 - 4 1759 89
          BC049112.1 - 4 1941 6415577 89
          BC054831.1 - 4 1740 6306945 89

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tubb4a

          Panther Classification:

          Molecular Function -

          tubulin

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule-based process
          cell projection organization
          protein polymerization
          nucleotide binding
          GTPase activity
          structural constituent of cytoskeleton
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259986 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259987 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259989 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259988 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P315795 M28730.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P315795 AK013717.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P315795 BC049112.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P315795 BC054831.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P315795 NM_009451.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P315795 M28730.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P315795 AK013717.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P315795 BC049112.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P315795 BC054831.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P315795 NM_009451.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 627732 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 324193 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 67912 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 555970 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 502117 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 166429 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120682 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P315795 M28730.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P315795 AK013717.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P315795 BC049112.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P315795 BC054831.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P315795 NM_009451.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P315795 M28730.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P315795 AK013717.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P315795 BC049112.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P315795 BC054831.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P315795 NM_009451.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315795 M28730.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315795 AK013717.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315795 BC049112.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315795 BC054831.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315795 NM_009451.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315795 M28730.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315795 AK013717.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315795 BC049112.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315795 BC054831.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P315795 NM_009451.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31260005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31260011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31260004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31260000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31260013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31260008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31260007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31260003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31260006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31259998 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120682 - -
          Whole Mouse Genome G4122A A_51_P315795 M28730.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315795 AK013717.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315795 BC049112.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315795 BC054831.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315795 NM_009451.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315795 M28730.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315795 AK013717.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315795 BC049112.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315795 BC054831.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P315795 NM_009451.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109324 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120682 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13589094 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13589098 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13270357 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13589097 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13589092 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13270356 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13589100 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259989 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259986 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259988 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31259987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31260028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31260014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31260004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31260026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31260002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31260009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31260000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31260020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31260013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31260025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31260021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31260011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31260006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31260005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31260017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31260007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31259893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13589094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31260000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13288131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31260006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31260017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31260014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13589093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31260013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31260005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13092807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13558192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31260002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13270356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31260007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13502658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13376945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31260008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31260004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13457947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13558206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13502655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13288132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13056493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31260019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13589089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31259982 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10452296 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452297 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452299 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452298 - -
          Affymetrix HT MG-430 PM Array Plate 1423221_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10452295 - -
          Mouse Genome 430 2.0 Array 1423221_PM_at - -
          Mouse Genome 430 2.0 Array 1423221_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22153_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22153_PM_at - -
          Affymetrix Murine Genome 6500A Array Msa12620_at - -
          Mouse Expression Set 430 1423221_PM_at - -
          Mouse Expression Set 430 1423221_at - -
          Murine Genome U74v2 Set 101419_PM_at - -
          Murine Genome U74v2 Set 101419_at - -
          Mouse Genome 430A 2.0 Array 1423221_at - -
          Mouse Genome 430A 2.0 Array 1423221_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423221_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423221_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 788091 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6180576 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006180576 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1236175 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236175 - -
          mouse-6 v1.1 (Illumina) scl49746.4_205-S - -
          Illumina Sentrix mouse-6.v1 bead array 6180576 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl49746.4_205-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6180576 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1236175 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1236175 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl49746.4_205-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline