Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00726619_s1
          See other MATR3 GE Assays ›
          Gene Symbol:
          Matr3
          Gene Name:
          matrin 3
          Gene Aliases:
          1110061A14Rik, 2810017I02Rik, AI841759, AW555618, D030046F20Rik, mKIAA0723
          Chromosome Location:
          Chr.18: 35562158 - 35592045 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00726619_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Matr3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010771.6 NP_034901.2 2 468 102
          GenBank mRNA AK087939.1 - 2 353 102
          AK129199.1 - 2 419 102
          AK135937.1 - 1 201 102
          AK161128.1 - 2 367 102
          AK164188.1 - 1 246 102
          BC029070.1 - 2 379 3989914 102
          BC090833.1 - 1 241 6854095 102
          CT010322.1 - 1 152 102

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Matr3

          Gene Ontology Categories:

          Function(s) Process(es)

          posttranscriptional regulation of gene expression
          nucleotide binding
          nucleic acid binding
          RNA binding
          zinc ion binding
          poly(A) RNA binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800002290.mm.1 NM_010771.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002290.mm.1 NM_010771.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002290.mm.1 TC1800000311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002289.mm.1 NM_010771.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002289.mm.1 NM_010771.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002289.mm.1 TC1800000311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002291.mm.1 NM_010771.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002291.mm.1 NM_010771.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002291.mm.1 TC1800000311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002280.mm.1 NM_010771.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002280.mm.1 NM_010771.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002280.mm.1 TC1800000311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002283.mm.1 NM_010771.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002283.mm.1 NM_010771.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002283.mm.1 TC1800000311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002292.mm.1 NM_010771.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002292.mm.1 NM_010771.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002292.mm.1 TC1800000311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002279.mm.1 NM_010771.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002279.mm.1 NM_010771.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002279.mm.1 TC1800000311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002285.mm.1 NM_010771.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002285.mm.1 NM_010771.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002285.mm.1 TC1800000311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002284.mm.1 NM_010771.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002284.mm.1 NM_010771.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002284.mm.1 TC1800000311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002288.mm.1 NM_010771.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002288.mm.1 NM_010771.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002288.mm.1 TC1800000311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002286.mm.1 NM_010771.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002286.mm.1 NM_010771.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002286.mm.1 TC1800000311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002287.mm.1 NM_010771.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002287.mm.1 NM_010771.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002287.mm.1 TC1800000311.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 610849 - -
          ABI Mouse Genome Survey Microarray 891132 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline