Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00727041_s1
          See other TIMM13 GE Assays ›
          Gene Symbol:
          Timm13
          Gene Name:
          translocase of inner mitochondrial membrane 13
          Gene Aliases:
          D10Ertd378e, Tim9, Timm9
          Chromosome Location:
          Chr.10: 80899450 - 80900969 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00727041_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Timm13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013895.4 NP_038923.1 3 849 67
          GenBank mRNA AK009041.1 - 3 643 67
          AK013230.1 - 3 647 67
          AK213641.1 - 1 259 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Timm13

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          protein transport
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286202 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286199 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286200 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286215 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286217 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286221 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286198 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286201 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286220 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286219 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338180 AK009041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338180 AK009152.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338180 AK013230.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338180 AK088984.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338180 BC011436.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338180 AK009041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338180 AK009152.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338180 AK013230.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338180 AK088984.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338180 BC011436.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102599 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338180 AK009041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338180 AK009152.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338180 AK013230.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338180 AK088984.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338180 BC011436.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338180 AK009041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338180 AK009152.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338180 AK013230.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338180 AK088984.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338180 BC011436.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P251325 AF144702.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27286232 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102599 - -
          Whole Mouse Genome G4122A A_52_P338180 AK009041.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338180 AK009152.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338180 AK013230.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338180 AK088984.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338180 BC011436.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338180 AK009041.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338180 AK009152.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338180 AK013230.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338180 AK088984.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338180 BC011436.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P251325 AF144702.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114632 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102599 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08440571 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08440582 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286219 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286200 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286199 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286215 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286202 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286221 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286220 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286201 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286198 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27286217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27286217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08440571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08440579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27286209 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10371008 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371007 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371011 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371009 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371010 - -
          Affymetrix HT MG-430 PM Array Plate 1455211_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10371006 - -
          Mouse Genome 430 2.0 Array 1455211_a_at - -
          Mouse Genome 430 2.0 Array 1455211_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 30055_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 30055_at - -
          Affymetrix Mouse Clariom S Array PSR1000018787.mm.1 NM_013895.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018787.mm.1 NM_013895.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018787.mm.1 TC1000002518.mm.1 MTA TC
          Mouse Expression Set 430 1417499_at - -
          Mouse Expression Set 430 1455211_PM_a_at - -
          Mouse Expression Set 430 1455211_a_at - -
          Mouse Expression Set 430 1417499_PM_at - -
          Murine Genome U74v2 Set 94012_PM_at - -
          Murine Genome U74v2 Set 94210_at - -
          Murine Genome U74v2 Set 94012_at - -
          Murine Genome U74v2 Set 94210_PM_at - -
          Mouse Genome 430A 2.0 Array 1417499_PM_at - -
          Mouse Genome 430A 2.0 Array 1455211_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1455211_a_at - -
          Mouse Genome 430A 2.0 Array 1417499_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 767713 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2450484 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002450484 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2693840 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2693840 - -
          mouse-6 v1.1 (Illumina) scl37716.2_16-S - -
          Illumina Sentrix mouse-6.v1 bead array 2450484 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37716.2_16-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2450484 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2693839 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2693839 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl37716.2_16-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline