Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00727224_s1
          See other TMEM37 GE Assays ›
          Gene Symbol:
          Tmem37
          Gene Name:
          transmembrane protein 37
          Gene Aliases:
          AI173373, AW540203, Pr, Pr1
          Chromosome Location:
          Chr.1: 120067377 - 120074069 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00727224_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem37
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019432.2 NP_062305.1 2 252 90
          GenBank mRNA AJ272046.1 - 2 204 90
          AK002424.1 - 2 271 90
          AK050134.1 - 2 252 90
          AK086093.1 - 2 257 90
          BC024613.1 - 2 248 4215775 90

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem37

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          calcium ion transport
          regulation of ion transmembrane transport
          calcium ion transmembrane transport
          voltage-gated ion channel activity
          calcium channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20560138 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20560140 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20560137 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401987 AK086093.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401987 BC024613.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401987 NM_019432.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401987 AK002424.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401987 AK050134.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401987 AK086093.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401987 BC024613.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401987 NM_019432.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401987 AK002424.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P401987 AK050134.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971679 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 288382 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375418 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 488773 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172390 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401987 AK086093.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401987 BC024613.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401987 NM_019432.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401987 AK002424.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401987 AK050134.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401987 AK086093.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401987 BC024613.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401987 NM_019432.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401987 AK002424.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P401987 AK050134.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401987 AK086093.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401987 BC024613.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401987 NM_019432.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401987 AK002424.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401987 AK050134.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401987 AK086093.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401987 BC024613.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401987 NM_019432.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401987 AK002424.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P401987 AK050134.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P25141 AK086093.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P25141 BC024613.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P25141 NM_019432.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P25141 AJ272046.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P25141 AK002424.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20560147 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04239142 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172390 - -
          Whole Mouse Genome G4122A A_51_P401987 AK086093.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401987 BC024613.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401987 NM_019432.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401987 AK002424.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401987 AK050134.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401987 AK086093.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401987 BC024613.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401987 NM_019432.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401987 AK002424.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P401987 AK050134.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P25141 AK086093.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P25141 BC024613.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P25141 NM_019432.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P25141 AJ272046.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P25141 AK002424.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114222 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172390 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00055097 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00000584 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00000588 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00000590 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20560137 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20560138 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20560140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20560179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560101 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00066379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00701559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00313007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00000579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00066391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00207317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00066383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00224551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00895559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00701563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00701560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560149 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00313011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01013715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00377839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560151 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00293434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00207315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00000585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00772106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00000586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20560166 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10357240 - -
          Affymetrix Mouse Gene 1.0 ST Array 10357241 - -
          Affymetrix HT MG-430 PM Array Plate 1417611_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10357239 - -
          Mouse Genome 430 2.0 Array 1417611_PM_at - -
          Mouse Genome 430 2.0 Array 1417611_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170706_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 170706_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100023832.mm.1 NM_019432.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023832.mm.1 NM_019432.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100023832.mm.1 TC0100002963.mm.1 MTA TC
          Mouse Expression Set 430 1417611_PM_at - -
          Mouse Expression Set 430 1417611_at - -
          Murine Genome U74v2 Set 95465_s_at - -
          Murine Genome U74v2 Set 95464_at - -
          Murine Genome U74v2 Set 95465_PM_s_at - -
          Murine Genome U74v2 Set 95464_PM_at - -
          Mouse Genome 430A 2.0 Array 1417611_PM_at - -
          Mouse Genome 430A 2.0 Array 1417611_at - -
          Affymetrix HT Mouse Genome 430A Array 1417611_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417611_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 580920 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1660170 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001660170 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230151 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230151 - -
          mouse-6 v1.1 (Illumina) scl16376.2_15-S - -
          Illumina Sentrix mouse-6.v1 bead array 1660170 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl16376.2_15-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1660170 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230151 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230151 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline