Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00727329_s1
          See other TUSC2 GE Assays ›
          Gene Symbol:
          Tusc2
          Gene Name:
          tumor suppressor candidate 2
          Gene Aliases:
          1190001E22Rik, AA407686, Fus1, Lgcc, PAP, Pdap2
          Chromosome Location:
          Chr.9: 107563255 - 107566108 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00727329_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tusc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019742.4 NP_062716.1 3 860 125
          GenBank mRNA AF123387.1 - 3 747 125
          AK082684.1 - 3 861 125

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tusc2

          Gene Ontology Categories:

          Function(s) Process(es)

          natural killer cell differentiation
          phagocytosis
          inflammatory response
          interleukin-15 production
          negative regulation of interleukin-17 production
          positive regulation of interleukin-10 production
          cell maturation
          defense response to Gram-negative bacterium
          regulation of mitochondrial membrane potential
          response to defense-related host reactive oxygen species production
          neutrophil mediated killing of gram-negative bacterium
          chemokine (C-C motif) ligand 5 production
          regulation of reactive oxygen species metabolic process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26809574 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26809576 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26809573 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P123354 BC048477.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P123354 NM_019742.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P123354 AF123387.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P123354 AK004430.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P123354 AK082684.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P123354 BC048477.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P123354 NM_019742.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P123354 AF123387.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P123354 AK004430.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P123354 AK082684.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 808511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 374159 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170251 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P123354 BC048477.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P123354 NM_019742.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P123354 AF123387.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P123354 AK004430.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P123354 AK082684.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P123354 BC048477.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P123354 NM_019742.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P123354 AF123387.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P123354 AK004430.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P123354 AK082684.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P372585 NM_019742.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P372585 AF123387.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P372585 AK082684.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P123354 BC048477.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P123354 NM_019742.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P123354 AF123387.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P123354 AK004430.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P123354 AK082684.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P123354 BC048477.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P123354 NM_019742.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P123354 AF123387.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P123354 AK004430.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P123354 AK082684.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809591 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809574 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170251 - -
          Whole Mouse Genome G4122A A_52_P123354 BC048477.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P123354 NM_019742.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P123354 AF123387.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P123354 AK004430.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P123354 AK082684.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P123354 BC048477.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P123354 NM_019742.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P123354 AF123387.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P123354 AK004430.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P123354 AK082684.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P372585 NM_019742.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P372585 AF123387.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P372585 AK082684.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111971 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104157 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170251 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07786614 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07786610 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07734400 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07734324 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07734336 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26809576 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26809573 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26809574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07734320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07734380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809567 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10588679 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588681 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588682 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588680 - -
          Affymetrix HT MG-430 PM Array Plate 1417593_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10588678 - -
          Affymetrix Mouse Gene 1.1 ST Array 10596671 - -
          Mouse Genome 430 2.0 Array 1417593_PM_at - -
          Mouse Genome 430 2.0 Array 1417593_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 80385_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 80385_at - -
          Mouse Expression Set 430 1417593_PM_at - -
          Mouse Expression Set 430 1417593_at - -
          Murine Genome U74v2 Set 160142_PM_at - -
          Murine Genome U74v2 Set 160142_at - -
          Mouse Genome 430A 2.0 Array 1417593_at - -
          Mouse Genome 430A 2.0 Array 1417593_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417593_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417593_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 586218 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1580253 - -
          Illumina mouse-6 v1.0 expression beadchip 1580253 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001580253 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2522023 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2522023 - -
          mouse-6 v1.1 (Illumina) scl36493.4_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 1580253 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36493.4_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1580253 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2834451 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2522023 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2522023 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2834451 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl36493.4_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0