| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26239786 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26239790 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26239789 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2041828 |
BC031357.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2041828 |
BC088749.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2041828 |
NM_023279.2 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2041828 |
AF312873.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
961286 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
654563 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
356691 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
528004 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
798004 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
894849 |
XM_001476668.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
10728 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
98243 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P139831 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P129850 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2041828 |
BC031357.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2041828 |
BC088749.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2041828 |
NM_023279.2 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2041828 |
AF312873.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P381381 |
BC031357.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P381381 |
BC088749.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P381381 |
AK051298.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P381381 |
AK082942.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P381381 |
AK149014.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239799 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239806 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239771 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239782 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239764 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239786 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239787 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239796 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239785 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239773 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239792 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239801 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239781 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239789 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239780 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239776 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239770 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239794 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239777 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239778 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239808 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239774 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239805 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239768 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239783 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239809 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239804 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P26239797 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P139831 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P129850 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P381381 |
BC031357.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P381381 |
BC088749.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P381381 |
AK051298.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P381381 |
AK082942.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P381381 |
AK149014.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P140941 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P129850 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P139831 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07097351 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07097360 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P07097356 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26239789 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26239790 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P26239786 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239733 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239799 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239738 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239783 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239862 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239811 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239821 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239831 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239813 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239858 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239826 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239759 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239830 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239750 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239787 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239866 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239846 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239804 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239785 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239809 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239746 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239755 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239819 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239792 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239801 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239794 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239740 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239736 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239805 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239815 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239782 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239753 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239756 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239789 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239839 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239859 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239745 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239796 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239761 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239749 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239762 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239827 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239872 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239752 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239824 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239864 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239870 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239734 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239832 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239748 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239797 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239731 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239806 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239833 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P26239863 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07179000 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239836 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06712265 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239797 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239785 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07083480 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239864 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239859 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239830 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239858 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239862 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239869 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239819 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239827 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239799 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239863 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07069108 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239790 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239846 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239866 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239796 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239809 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239870 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239872 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239805 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239813 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239792 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239860 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06826974 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239801 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07335604 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239826 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239815 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P07083481 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06760227 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06826981 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239783 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239811 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06826976 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239832 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239794 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239817 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239782 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239806 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06867757 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P26239857 |
- |
- |