Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00728347_s1
          See other NGRN GE Assays ›
          Gene Symbol:
          Ngrn
          Gene Name:
          neugrin, neurite outgrowth associated
          Gene Aliases:
          AW552001
          Chromosome Location:
          Chr.7: 80261215 - 80265378 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00728347_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ngrn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031375.4 NP_113552.3 3 862 102
          NR_028053.1 2 1259 102
          GenBank mRNA AB047544.1 - 3 888 102
          AK083330.1 - 2 1255 102
          AK144756.1 - 3 858 102
          AK146059.1 - 2 1237 102
          BC004034.1 - 849 3495639 102

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ngrn

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          nervous system development
          biological_process
          cell differentiation
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25265629 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25265630 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003133 AK083330.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003133 AK144756.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003133 NM_031375.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003133 NR_028053.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003133 AB047544.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487203 AK088787.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487203 AK145409.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487203 AK146820.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487203 AK161654.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487203 AK168702.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487203 AK168981.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487203 BC028849.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487203 BC032291.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487203 BC086664.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487203 NM_001013378.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 467558 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 467795 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 655613 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 340712 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 690944 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636572 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 805967 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 310869 AB008910.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 310869 BC049616.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 310869 BC115665.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 310869 BC115666.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 310869 NM_009355.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 141365 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 201549 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 694057 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 518171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 89721 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114790 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108594 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137475 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107160 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112878 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003133 AK083330.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003133 AK144756.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003133 NM_031375.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003133 NR_028053.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003133 AB047544.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25265648 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114790 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06256205 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01558752 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06256434 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01558774 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01558737 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107160 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01558713 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01558705 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01558613 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01558761 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06256224 - -
          Whole Mouse Genome G4122A A_51_P393999 CJ154012.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393999 AK083330.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393999 AK144756.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393999 AK146059.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393999 BC004034.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P393998 AB047544.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129855 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107160 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06256434 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114790 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01558761 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06556669 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06556662 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25265630 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25265629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25265702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25267307 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25267299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25267344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06294151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05978361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25267305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25267324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25267306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25267351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25267329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25267346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06450607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06371297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06049771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06231928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06556669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05978357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06385065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25267320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25267327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05914664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06034882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25265631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25267317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06411924 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10554418 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554417 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554416 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554414 - -
          Affymetrix Mouse Gene 1.0 ST Array 10554415 - -
          Affymetrix HT MG-430 PM Array Plate 1416709_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10554413 - -
          Mouse Genome 430 2.0 Array 1416709_a_at - -
          Mouse Genome 430 2.0 Array 1416709_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83485_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 83485_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700010087.mm.1 TC0700001149.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010084.mm.1 TC0700001149.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700010084.mm.1 NM_031375.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010084.mm.1 NM_031375.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010084.mm.1 NR_028053.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700010084.mm.1 NR_028053.1 Refseq
          Mouse Expression Set 430 1416709_PM_a_at - -
          Mouse Expression Set 430 1416709_a_at - -
          Murine Genome U74v2 Set 103875_PM_at - -
          Murine Genome U74v2 Set 103875_at - -
          Mouse Genome 430A 2.0 Array 1416709_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1416709_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416709_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1416709_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 820953 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4210390 - -
          Illumina mouse-6 v1.0 expression beadchip 4210390 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004210390 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2674296 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2674296 - -
          mouse-6 v1.1 (Illumina) scl083485.3_53-S - -
          Illumina Sentrix mouse-6.v1 bead array 4210390 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl083485.3_53-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4210390 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2674296 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2674296 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline