Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00728712_s1
          See other ZFP958 GE Assays ›
          Gene Symbol:
          Zfp958
          Gene Name:
          zinc finger protein 958
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 4613170 - 4630231 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00728712_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp958
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145591.4 NP_663566.1 4 2034 120
          GenBank mRNA AK037083.1 - 4 2036 121

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp958

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2068429 BC063063.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2068429 AK013418.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098573 AK037083.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2098573 NM_145591.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 158146 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 797438 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 311521 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 149366 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377059 NM_009154.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377059 X97817.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 459032 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 59560 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 166317 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125829 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165135 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152416 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2068429 BC063063.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2068429 AK013418.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098573 AK037083.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2098573 NM_145591.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25618110 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152416 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165135 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125829 - -
          Whole Mouse Genome G4122A A_52_P304281 BC138223.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P304281 BC145996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P304281 AK037083.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P304281 AK037596.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P304281 BC003267.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122273 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136658 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165135 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152416 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125829 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07296135 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07304890 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07296138 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07296131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25618159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06925834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07337144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07296131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07383717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07383723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06686747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06741849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07304890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07296137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25618137 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10569988 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569990 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569989 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569987 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569991 - -
          Affymetrix Mouse Gene 1.0 ST Array 10569986 - -
          Affymetrix HT MG-430 PM Array Plate 1460369_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10569985 - -
          Mouse Genome 430 2.0 Array 1460369_at - -
          Mouse Genome 430 2.0 Array 1460369_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233987_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233987_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800000491.mm.1 NM_145591.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000491.mm.1 NM_145591.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000491.mm.1 TC0800000039.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800000490.mm.1 NM_145591.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000490.mm.1 NM_145591.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000490.mm.1 TC0800000039.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800000492.mm.1 NM_145591.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000492.mm.1 NM_145591.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800000492.mm.1 TC0800000039.mm.1 MTA TC
          Mouse Expression Set 430 1460369_at - -
          Mouse Expression Set 430 1460369_PM_at - -
          Mouse Genome 430A 2.0 Array 1460369_at - -
          Mouse Genome 430A 2.0 Array 1460369_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460369_at - -
          Affymetrix HT Mouse Genome 430A Array 1460369_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 587884 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1780079 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001780079 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222712 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222712 - -
          mouse-6 v1.1 (Illumina) scl34111.4_462-S - -
          Illumina Sentrix mouse-6.v1 bead array 1780079 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34111.4_462-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1780079 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1222712 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222712 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0