Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00728872_s1
          See other OLFR1279 GE Assays ›
          Gene Symbol:
          Olfr1279
          Gene Name:
          olfactory receptor 1279
          Gene Aliases:
          MOR245-12
          Chromosome Location:
          Chr.2: 111306201 - 111307142 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00728872_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1279
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146393.1 NP_666505.1 1 852 106
          GenBank mRNA BC113137.1 - 1 852 40042162 106

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1279

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2124946 BC113137.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2124946 NM_146393.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2124946 BC113137.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2124946 NM_146393.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P369451 BC113137.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P369451 NM_146393.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21524768 - -
          Whole Mouse Genome G4122A A_52_P369451 BC113137.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P369451 NM_146393.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127993 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100543 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01898490 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01898501 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01898484 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01898509 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01898505 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01898489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21524788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01618107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01618106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21524807 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10474484 - -
          Affymetrix Mouse Gene 1.1 ST Array 10474483 - -
          Affymetrix Mouse Clariom S Array PSR0200012259.mm.1 NM_146393.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200012259.mm.1 NM_146393.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200012259.mm.1 TC0200001641.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 403206 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2480136 - -
          Illumina mouse-6 v1.0 expression beadchip 2480136 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002480136 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230736 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230736 - -
          mouse-6 v1.1 (Illumina) scl20495.1.1_330-S - -
          Illumina Sentrix mouse-6.v1 bead array 2480136 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl20495.1.1_330-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2480136 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230736 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230736 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline