Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00728958_s1
          See other OLFR1289 GE Assays ›
          Gene Symbol:
          Olfr1289
          Gene Name:
          olfactory receptor 1289
          Gene Aliases:
          MOR245-6
          Chromosome Location:
          Chr.2: 111483432 - 111484328 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00728958_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1289
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146404.1 NP_666516.1 1 256 125
          GenBank mRNA BC120736.1 - 1 338 40129659 125
          BC137761.1 - 1 339 8860780 125

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1289

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137536 BC120736.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137536 BC137761.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2137536 NM_146404.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107643 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2137536 BC120736.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2137536 BC137761.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2137536 NM_146404.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P250887 BC120736.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P250887 BC137761.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P250887 NM_146404.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21525510 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107643 - -
          Whole Mouse Genome G4122A A_51_P250887 BC120736.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P250887 BC137761.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P250887 NM_146404.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115647 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107643 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02062087 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02062083 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02062054 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02062036 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02062037 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02062045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21525454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01537651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01807751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02062085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02062072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21525511 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10474505 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474502 - -
          Affymetrix Mouse Gene 1.0 ST Array 10474503 - -
          Affymetrix Mouse Gene 1.1 ST Array 10474504 - -
          Affymetrix Mouse Gene 1.1 ST Array 10474501 - -
          Affymetrix Mouse Clariom S Array PSR0200012278.mm.1 TC0200001649.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200012279.mm.1 NM_146404.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200012279.mm.1 NM_146404.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200012279.mm.1 TC0200001649.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200012280.mm.1 TC0200001649.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 494189 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4670446 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004670446 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1249853 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249853 - -
          mouse-6 v1.1 (Illumina) scl20494.3.191_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 4670446 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl20494.3.191_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4670446 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1249853 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2866545 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1249853 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2866545 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline